Skip to Content
Merck
All Photos(1)

Key Documents

93443

Sigma-Aldrich

Triton X-100 solution

Triton™ X-100 solution
1 of 2 reviewers received a sample product or took part in a promotion

BioUltra, for molecular biology, ~10% in H2O

Synonym(s):

Triton X-100, 4-(1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol solution, t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10
CAS Number:
Beilstein:
2315025
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

grade

for molecular biology

Quality Level

description

non-ionic

product line

BioUltra

form

liquid

mol wt

micellar avg mol wt 80,000
average mol wt 625

concentration

~10% in H2O

aggregation number

100-155

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

refractive index

n20/D 1.349±0.002

pH

6.5-8.5 (25 °C)

CMC

0.2-0.9 mM (20-25°C)
0.23 mM

transition temp

cloud point 65 °C
pour point ~7 °C

density

1.01 g/mL at 20 °C
1.070 g/cm3

cation traces

Al: ≤1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤100 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤100 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

absorption

cut-off at 310 nm

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

HLB

13.5

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triton X-100 is a common non-ionic surfactant and emulsifier which is often used in biochemical applications to solubilize proteins. It is considered a comparatively mild detergent, non-denaturing, and is reported in numerous references as a routinely added reagent. It is utilized for lysing cells to extract protein and cellular organelles. It can also permeabilize the living cell membrane for transfection.[1][2]

Application

Triton X-100 has been used:
  • for permeabilization of cells before techniques like indirect immunofluorescence and flow cytometry[3][4]
  • As a component of lysis buffer used for homogenization of cells[3][4][5]
  • As a component of blocking buffer in immunofluorescence staining[6]
  • For the preparation of cells in double fluorescence staining[4]

Biochem/physiol Actions

Widely used non-ionic surfactant for recovery of membrane components under mild non-denaturing conditions.

Features and Benefits

  • Suitable for Molecular Biology and Biochemical Research
  • Free from DNase, RNase, NICKase and protease
  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications.
  • High purity product for research applications
  • Tested for the presence of Anionic and Cationic traces

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

483.8 °F - closed cup

Flash Point(C)

251 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Melittin exerts an antitumor effect on non small cell lung cancer cells.
Zhang S F and Chen Z
Molecular Medicine Reports, 16(3), 3581-3586 (2017)
The effect of artificial shrinkage and assisted hatching on the development of mouse blastocysts and cell number after vitrification.
Kim H J, et al.
Clinical and Experimental Reproductive Medicine, 42(3), 94-100 (2015)
Creation of an injectable in situ gelling native extracellular matrix for nucleus pulposus tissue engineering.
Wachs R A, et al.
The Spine Journal : Official Journal of the North American Spine Society, 17(3), 435-444 (2017)
Alena Balashova et al.
Frontiers in neuroscience, 13, 1033-1033 (2019-10-22)
It is well-known that hyaluronic acid (HA) as a component of brain extracellular matrix (ECM) plays a pivotal role in the nervous system and is involved in synaptic plasticity changes in vascular cognitive impairment and dementia. HA breakdown is a
Oxysterols in adipose tissue-derived mesenchymal stem cell proliferation and death.
Silva S F, et al.
The Journal of Steroid Biochemistry and Molecular Biology, 169, 164-175 (2017)

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the concentration of X100, Triton® X-100?

    1 answer
    1. The concentration of this product is approximately 10%. The tolerance is < 1% with a resulting general range of 9.01 - 10.99%. This information is not a specification nor is it reported on a lot-specific basis.

      Helpful?

Reviews

1 of 2 reviewers received a sample product or took part in a promotion

Active Filters

  1. USA
    • Review 1
    • Votes 0
    5 out of 5 stars.

    Great Customer Service Staff

    Item came quick and with no issues- stable packaging and as advertised. Real star was the customer service staff. Quick, professional, and friendly- able to help me out along the whole process.

    Helpful?

  2. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service