Skip to Content
Merck
All Photos(1)

Key Documents

X165

Sigma-Aldrich

Triton X-165 solution

Non-ionic, CMC-0.43 mM, suitable for electrophoresis, cloud point >100 °C

Synonym(s):

Ethoxylated octylphenol, Octylphenol ethoxylate, Polyethylene glycol mono(octylphenyl) ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.21

Quality Level

description

non-ionic

mol wt

911 g/mol

technique(s)

electrophoresis: suitable
protein quantification: suitable

CMC

0.43 mM

transition temp

cloud point >100 °C

HLB

15.5

SMILES string

O(CCOc1ccc(cc1)C(CC(C)(C)C)(C)C)CCO

InChI

1S/C18H30O3/c1-17(2,3)14-18(4,5)15-6-8-16(9-7-15)21-13-12-20-11-10-19/h6-9,19H,10-14H2,1-5H3

InChI key

LBCZOTMMGHGTPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triton X-165 is a non-ionic surfactant that is highly versatile and can be used in a range of laboratory applications. It is soluble in water and miscible in most polar organic solvents, but insoluble in aliphatic and aromatic hydrocarbons. This makes it an excellent choice for preparing iodophors and dispersions. Additionally, Triton X-165 is highly chemically stable in most acidic and alkaline solutions, which makes it ideal for researchers who need a reliable and effective nonionic surfactant. If you need a high-quality nonionic surfactant for your laboratory applications, Triton X-165 is an excellent choice. Its proven versatility, solubility, and chemical stability make it a valuable tool for researchers in a range of fields, including biochemistry, biophysics, and molecular biology.

Application

Triton X-165 solution is suitable for use in:

  • determining its effect on the stability of the multi-walled carbon nanotubes (MWNT) suspension in isopropyl alcohol
  • studying its surface tension-dependent toxicity on the green peach aphid, Myzus persicae to determine the mechanism of surfactant action
  • paints and coatings, agrochemicals, fiber lubricants, emulsion polymerization

Features and Benefits

  • Excellent steric and freeze/thaw stabilizer
  • Excellent for aromatic, chlorinated, and other compounds that are difficult to emulsify
  • Offers good solubility in the presence of salts or electrolytes and caustic solutions
  • Effective at high temperatures
  • Compatible with anionic, cationic, and other nonionic surfactants

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of surfactant addition on the MWNT suspenzions stability in organic solvents
Gigiberiya VA et al.
Journal of Separation Science (2011)
Surface tension-dependent surfactant toxicity on the green peach aphid, Myzus persicae (Sulzer)(Hemiptera: Aphididae)
IMAI, et al.
Journal of Separation Science, 29(3), 389-393 (1994)
Effect of surfactant addition on the MWNT suspenzions stability in organic solvents
Gigiberiya VA et al.
Journal of Separation Science (2011)
Russell M Morphew et al.
Veterinary parasitology, 191(3-4), 379-385 (2012-10-02)
Fasciola hepatica NP-40 released protein extract (FhNPE) exhibits potent Th1 immunosuppressive properties in vitro and in vivo. However, the protein composition of this active fraction, responsible for Th1 immune modulatory activity, has yet to be resolved. Therefore, FhNPE, a Nonidet
Halil İbrahim Ulusoy et al.
Talanta, 88, 516-523 (2012-01-24)
A new micelle-mediated separation and preconcentration method was developed for ultra-trace quantities of mercury ions prior to spectrophotometric determination. The method is based on cloud point extraction (CPE) of Hg(II) ions with polyethylene glycol tert-octylphenyl ether (Triton X-114) in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service