Merck
A general analytical platform and strategy in search for illegal drugs.
Journal of pharmaceutical and biomedical analysis (2014-08-30)
Monika Johansson, Dick Fransson, Torgny Rundlöf, Ngoc-Hang Huynh, Torbjörn Arvidsson
ABSTRACT
An effective screening procedure to identify and quantify active pharmaceutical substances in suspected illegal medicinal products is described. The analytical platform, consisting of accurate mass determination with liquid chromatography time-of-flight mass spectrometry (LC-QTOF-MS) in combination with nuclear magnetic resonance (NMR) spectroscopy provides an excellent analytical tool to screen for unknowns in medicinal products, food supplements and herbal formulations. This analytical approach has been successfully applied to analyze thousands of samples. The general screening method usually starts with a methanol extraction of tablets/capsules followed by liquid chromatographic separation on a Halo Phenyl-Hexyl column (2.7μm; 100mm×2.1mm) using an acetonitrile/0.1% formic acid gradient as eluent. The accurate mass of peaks of interest was recorded and a search made against an in-house database containing approximately 4200 substances, mostly pharmaceutical compounds. The search could be general or tailored against different classes of compounds. Hits were confirmed by analyzing a reference substance and/or by NMR. Quantification was normally performed with quantitative NMR (qNMR) spectroscopy. Applications for weight-loss substances like sibutramine and orlistat, sexual potency enhancement (PDE-5 inhibitors), and analgesic drugs are presented in this study. We have also identified prostaglandin analogues in eyelash growth serum, exemplified by isopropyl cloprostenate and bimatoprost. For creams and ointments, matrix solid-phase dispersion (MSPD) was found to give a clean extracts with high recovery prior to LC-MS analyses. The structural elucidation of cetilistat, a new weight-loss substance recently found in illegal medicines purchased over the Internet, is also presented.
Product Description
Triamcinolone acetonide, European Pharmacopoeia (EP) Reference Standard
USP
Triamcinolone acetonide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
3,4,5-Trichloropyridine, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Supelco
Methanol, analytical standard
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Butadiene sulfone, 98%
Supelco
Triamcinolone acetonide, analytical standard
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material