Skip to Content
Merck
All Photos(3)

Key Documents

T6501

Supelco

Triamcinolone acetonide

analytical standard

Synonym(s):

9α-Fluoro-11β,16α,17α,21-tetrahydroxy-1,4-pregnadiene-3,20-dione 16,17-acetonide, 9α-Fluoro-16α-hydroxyprednisolone 16α,17α-acetonide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31FO6
CAS Number:
Molecular Weight:
434.50
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥99%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1(F)[C@@H](O)C[C@@]4(C)[C@@]2([H])C[C@H]5OC(C)(C)O[C@@]45C(=O)CO

InChI

1S/C24H31FO6/c1-20(2)30-19-10-16-15-6-5-13-9-14(27)7-8-21(13,3)23(15,25)17(28)11-22(16,4)24(19,31-20)18(29)12-26/h7-9,15-17,19,26,28H,5-6,10-12H2,1-4H3/t15-,16-,17-,19+,21-,22-,23-,24+/m0/s1

InChI key

YNDXUCZADRHECN-JNQJZLCISA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie-Hélène Errera et al.
Survey of ophthalmology, 58(2), 127-142 (2013-02-16)
Pregnancy-associated retinal diseases are conditions that may occur uniquely in pregnancy or, more commonly, general conditions that may worsen or alter during pregnancy as a result of hematologic, hormonal, metabolic, cardiovascular, and immunologic changes. Diabetic retinopathy (DR) is by far
Tzu-Yun Kao et al.
American journal of ophthalmology, 156(1), 149-156 (2013-05-03)
To investigate the autofluorescence patterns of sealed macular holes and determine the correlations of the postoperative changes in spectral-domain optical coherence tomography and fundus autofluorescence (FAF) with the best-corrected visual acuity (BCVA) of patients with sealed idiopathic macular holes. Retrospective
Miriam Englander et al.
The British journal of ophthalmology, 97(4), 460-465 (2013-02-08)
To report the incidence rate of acute postoperative endophthalmitis secondary to therapeutic intravitreal injections. A retrospective review of all consecutive eyes after intravitreal injections was performed at the Massachusetts Eye and Ear Infirmary, Boston, from 1 January 2007 to 31
Shivani Kasbekar et al.
Journal of cataract and refractive surgery, 39(3), 414-418 (2013-01-23)
To compare the clinical outcomes in patients who had triamcinolone acetate-assisted anterior vitrectomy and patients who had anterior vitrectomy without triamcinolone acetate after phacoemulsification complicated by posterior capsule rupture and vitreous loss. Arrowe Park Hospital, Wirral, United Kingdom. Retrospective consecutive
Brian C Gilger et al.
Investigative ophthalmology & visual science, 54(4), 2483-2492 (2013-03-28)
To evaluate the effect of triamcinolone acetonide (TA) administered into the suprachoroidal space (SCS) using a microneedle and compare it with intravitreal (IVT) TA injections in a porcine model of acute posterior segment inflammation. An IVT injection of balanced salt

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service