Skip to Content
Merck
All Photos(1)

Key Documents

PHR1320

Supelco

Residual Solvent - Acetonitrile(solution in DMSO)

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Acetonitrile, ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1601340

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)

CofA

current certificate can be downloaded

autoignition temp.

973 °F

expl. lim.

16 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

density

0.786 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-30°C

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Residual Solvent - Acetonitrile is a CRM and Secondary Pharmaceutical Reference Standard, traceable and qualified against the corresponding Pharmacopeial standards. Sigma-Aldrich offers Certified Reference Materials (CRMs) for the analysis of Residual Solvents. A comprehensive Certificate of Analysis offers details of the methodology and results of the qualification.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA0679 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elke H J Krekels et al.
Clinical pharmacokinetics, 53(6), 553-563 (2014-02-06)
From a previously validated paediatric population pharmacokinetic model, it was derived that non-linear morphine maintenance doses of 5 μg/kg(1.5)/h, with a 50 % dose reduction in neonates with a postnatal age (PNA) <10 days, yield similar morphine and metabolite concentrations
N Sorvillo et al.
Journal of thrombosis and haemostasis : JTH, 12(5), 670-679 (2014-07-01)
Acquired deficiency of ADAMTS13 causes a rare and life-threatening disorder called thrombotic thrombocytopenic purpura (TTP). Several studies have shown that aberrant glycosylation can play an important role in the pathogenesis of autoimmune diseases.N-linked glycosylation and putative O-fucosylation sites have been
Hélène Hall et al.
Brain : a journal of neurology, 137(Pt 9), 2493-2508 (2014-07-27)
The neuropathological substrate of dementia in patients with Parkinson's disease is still under debate, particularly in patients with insufficient alternate neuropathology for other degenerative dementias. In patients with pure Lewy body Parkinson's disease, previous post-mortem studies have shown that dopaminergic
Divya Subramonian et al.
Journal of proteome research, 13(9), 3905-3918 (2014-07-30)
SUMOylation is an essential posttranslational modification and regulates many cellular processes. Dysregulation of SUMOylation plays a critical role in metastasis, yet how its perturbation affects this lethal process of cancer is not well understood. We found that SUMO-2/3 modification is
Changting Xiao et al.
Diabetes, 63(7), 2394-2401 (2014-03-04)
The dipeptidyl peptidase-4 inhibitor sitagliptin, an antidiabetic agent, which lowers blood glucose levels, also reduces postprandial lipid excursion after a mixed meal. The underlying mechanism of this effect, however, is not clear. This study examined the production and clearance of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service