Skip to Content
Merck
All Photos(3)

Key Documents

A1542

Sigma-Aldrich

Ammonium acetate

for molecular biology, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2NH4
CAS Number:
Molecular Weight:
77.08
Beilstein:
4186741
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
39021908
PubChem Substance ID:
NACRES:
NA.25

grade

Molecular Biology
for molecular biology

Quality Level

Assay

≥98%

form

solid

pH

6.5-7.5 (25 °C, 77.1 g/L)

mp

110-112 °C (dec.) (lit.)

solubility

water: 1 g/mL, clear, colorless to faintly yellow

anion traces

chloride (Cl-): ≤5 ppm

cation traces

Fe: ≤5 ppm
heavy metals (as Pb): ≤5 ppm

application(s)

diagnostic assay manufacturing

foreign activity

DNase, RNase and protease, none detected

storage temp.

2-8°C

SMILES string

N.CC(O)=O

InChI

1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3

InChI key

USFZMSVCRYTOJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • A green analytical method for the simultaneous determination of 17 perfluoroalkyl substances (PFAS) in human serum and semen by ultra-performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS).: This study highlights the use of ammonium acetate in chromatography as a part of a green analytical method, enhancing the detection sensitivity and specificity for environmental contaminants in biological samples (Di Giorgi et al., 2024).

  • Innovation of 6-sulfonamide-2H-chromene derivatives as antidiabetic agents targeting α-amylase, α-glycosidase, and PPAR-γ inhibitors with in silico molecular docking simulation.: Ammonium acetate is used in the synthesis and analysis of potential antidiabetic agents, demonstrating its utility in drug discovery and pharmaceutical chemistry (Thabet et al., 2024).

  • Regulation of Tetramethylpyrazine Formation by the Phenolics-Fenton Coupled Redox Cycling System.: Explores the role of ammonium acetate in regulating chemical reactions within food chemistry, impacting flavor development and food safety (Xu et al., 2024).

  • Simultaneous determination of unecritinib (TQ-B3101) and its active metabolite crizotinib in rat plasma by LC-MS/MS:An application to pharmacokinetic studies.: Utilizes ammonium acetate in liquid chromatography-mass spectrometry to enhance the analysis of pharmaceutical compounds, crucial for understanding drug metabolism and pharmacokinetics (Wang et al., 2024).

  • Combination of online hollow fiber liquid phase microextraction with smartphone-based sensing for in situ formaldehyde assay in fabric and wastewater samples.: Demonstrates an innovative use of ammonium acetate in environmental monitoring, combining it with new technologies to enhance detection capabilities in complex samples (Javadian et al., 2024).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loic Auderset et al.
Frontiers in cell and developmental biology, 8, 564351-564351 (2020-12-08)
Low-density lipoprotein receptor-related protein 1 (LRP1) is a large, endocytic cell surface receptor that is highly expressed by oligodendrocyte progenitor cells (OPCs) and LRP1 expression is rapidly downregulated as OPCs differentiate into oligodendrocytes (OLs). We report that the conditional deletion
Jean-François Daudelin et al.
Veterinary research, 42, 69-69 (2011-05-25)
This study evaluated the effect of the probiotics Pediococcus acidilactici and Saccharomyces cerevisiae boulardii on the intestinal colonization of O149 enterotoxigenic Escherichia coli harbouring the F4 (K88) fimbriae (ETEC F4) and on the expression of ileal cytokines in weaned pigs.
S Baron-Cohen et al.
Molecular psychiatry, 20(3), 369-376 (2014-06-04)
Autism affects males more than females, giving rise to the idea that the influence of steroid hormones on early fetal brain development may be one important early biological risk factor. Utilizing the Danish Historic Birth Cohort and Danish Psychiatric Central
Nazish Ahmed et al.
Tissue engineering. Part A, 20(15-16), 2224-2233 (2014-03-13)
One of the factors preventing clinical application of regenerative medicine to degenerative cartilage diseases is a suitable source of cells. Chondrocytes, the only cell type of cartilage, grown in vitro under culture conditions to expand cell numbers lose their phenotype
Martijs J Jonker et al.
Nucleic acids research, 42(11), e94-e94 (2014-04-29)
Structural variations in genomes are commonly studied by (micro)array-based comparative genomic hybridization. The data analysis methods to infer copy number variation in model organisms (human, mouse) are established. In principle, the procedures are based on signal ratios between test and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service