Skip to Content
Merck
All Photos(1)

Key Documents

T1900000

Triamcinolone acetonide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

9α-Fluoro-11β,16α,17α,21-tetrahydroxy-1,4-pregnadiene-3,20-dione 16,17-acetonide, 9α-Fluoro-16α-hydroxyprednisolone 16α,17α-acetonide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31FO6
CAS Number:
Molecular Weight:
434.50
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

triamcinolone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1(F)[C@@H](O)C[C@@]4(C)[C@@]2([H])C[C@H]5OC(C)(C)O[C@@]45C(=O)CO

InChI

1S/C24H31FO6/c1-20(2)30-19-10-16-15-6-5-13-9-14(27)7-8-21(13,3)23(15,25)17(28)11-22(16,4)24(19,31-20)18(29)12-26/h7-9,15-17,19,26,28H,5-6,10-12H2,1-4H3/t15-,16-,17-,19+,21-,22-,23-,24+/m0/s1

InChI key

YNDXUCZADRHECN-JNQJZLCISA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Triamcinolone acetonide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie-Hélène Errera et al.
Survey of ophthalmology, 58(2), 127-142 (2013-02-16)
Pregnancy-associated retinal diseases are conditions that may occur uniquely in pregnancy or, more commonly, general conditions that may worsen or alter during pregnancy as a result of hematologic, hormonal, metabolic, cardiovascular, and immunologic changes. Diabetic retinopathy (DR) is by far
Tzu-Yun Kao et al.
American journal of ophthalmology, 156(1), 149-156 (2013-05-03)
To investigate the autofluorescence patterns of sealed macular holes and determine the correlations of the postoperative changes in spectral-domain optical coherence tomography and fundus autofluorescence (FAF) with the best-corrected visual acuity (BCVA) of patients with sealed idiopathic macular holes. Retrospective
Miriam Englander et al.
The British journal of ophthalmology, 97(4), 460-465 (2013-02-08)
To report the incidence rate of acute postoperative endophthalmitis secondary to therapeutic intravitreal injections. A retrospective review of all consecutive eyes after intravitreal injections was performed at the Massachusetts Eye and Ear Infirmary, Boston, from 1 January 2007 to 31
Arthur Geltzer et al.
The Cochrane database of systematic reviews, 1(1), CD005022-CD005022 (2013-02-27)
Neovascular age-related macular degeneration (AMD) is associated with rapid vision loss due to choroidal neovascularization (CNV), leakage, and scarring. Steroids have gained attention in their role for the treatment of neovascular AMD for their antiangiogenic and anti-inflammatory properties. This review
Brian C Gilger et al.
Investigative ophthalmology & visual science, 54(4), 2483-2492 (2013-03-28)
To evaluate the effect of triamcinolone acetonide (TA) administered into the suprachoroidal space (SCS) using a microneedle and compare it with intravitreal (IVT) TA injections in a porcine model of acute posterior segment inflammation. An IVT injection of balanced salt

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service