Skip to Content
Merck
All Photos(1)

Documents

S2454

Millipore

Seppro Ammonium Bicarbonate Buffer

Synonym(s):

ABC buffer, Ammonium hydrogen carbonate, Monoammonium carbonate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.32

form

solution

Quality Level

concentration

2 M

technique(s)

protein expression: suitable

color

clear colorless

application(s)

life science and biopharma

storage temp.

room temp

General description

Seppro® Ammonium Bicarbonate Buffer is an effective buffer for protein digestion and deglycosylation reaction. It may be used to coat 2D and 3D electrophoretic gel pieces during digestion of proteins and proteomes. The ammonium bicarbonate buffer also provides moisture during enzymatic cleavage. Ammonium bicarbonate buffer regulates the microenvironmental pH of immobilized cephalosporin C acylase (CCA) for the catalysis of cephalosporin C (CPC) in a batch stirred tank reactor (STR).

Application

Ammonium bicarbonate buffer has been used as a component of cell lysis/protein extraction buffer for proteomic sample preparation.

Legal Information

6538 is a trademark of American Type Culture Collection
Seppro is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrej Shevchenko et al.
Nature protocols, 1(6), 2856-2860 (2007-04-05)
In-gel digestion of proteins isolated by gel electrophoresis is a cornerstone of mass spectrometry (MS)-driven proteomics. The 10-year-old recipe by Shevchenko et al. has been optimized to increase the speed and sensitivity of analysis. The protocol is for the in-gel
Yanhong Chang et al.
Biotechnology progress, 35(5), e2846-e2846 (2019-05-18)
In a stirred tank reactor, during catalysis with immobilized cephalosporin C acylase (CCA), the microenvironmental pH dropped to 7.2 in a nonbuffered system (with the pH maintained at 8.5 by adding alkali) due to the existence of diffusional resistance. Moreover
Vimal K Balasubramanian et al.
Current protocols, 1(5), e153-e153 (2021-05-28)
Plant organs and tissues contain multiple cell types, which are well organized in 3-dimensional structure to efficiently perform physiological functions such as homeostasis and response to environmental perturbation and pathogen infection. It is critically important to perform molecular measurements at
Emily S Boja et al.
The Journal of biological chemistry, 278(36), 34189-34202 (2003-06-12)
The zona pellucida is an extracellular matrix consisting of three glycoproteins that surrounds mammalian eggs and mediates fertilization. The primary structures of mouse ZP1, ZP2, and ZP3 have been deduced from cDNA. Each has a predicted signal peptide and a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service