Skip to Content
Merck
All Photos(3)

Documents

379999

Sigma-Aldrich

Ammonium carbonate

99.999% trace metals basis

Synonym(s):

Hartshorn salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2CO3
CAS Number:
Molecular Weight:
96.09
Beilstein:
4253487
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38020208
PubChem Substance ID:
NACRES:
NA.55

grade

for inorganic trace analysis

vapor density

2.7 (vs air)

vapor pressure

1,013.25 hPa °F

description

mixture of variable proportions of ammonium bicarbonate and ammonium carbamate

Assay

99.999% trace metals basis

form

powder

solubility

soluble (Water: slightly soluble)

SMILES string

N.N.OC(O)=O

InChI

1S/CH2O3.2H3N/c2-1(3)4;;/h(H2,2,3,4);2*1H3

InChI key

PRKQVKDSMLBJBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium carbonate is an ammonium salt consisting varying amounts of ammonium bicarbonate and ammonium carbamate. The enthalpy of absorption of CO2 in aqueous ammonium carbonate has been determined using reaction calorimeter.

Application

Ammonium carbonate may be used:
  • As a dispersant for SiO2 nano particles for dynamic light scattering, Z-potential, and centrifugal liquid sedimentation measurements.
  • As an eluent in the separation of silica nanoparticles by asymmetric flow field-flow fractionation (AF4) coupled to inductively coupled plasma mass spectrometry (ICPMS) method.
  • In the synthesis of ZnO nanoparticles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nano ZnO synthesis by modified sol gel method and its application in heterogeneous photocatalytic removal of phenol from water.
Hayat K, et al.
Applied Catalysis A: General, 393(1), 122-129 (2011)
Adel Achouba et al.
Chemosphere, 229, 549-558 (2019-05-18)
Nunavimmiut (Inuit of Nunavik, Northern Quebec, Canada) exhibit a high selenium (Se) status because of their frequent consumption of marine mammal foods. Indirect evidence from our previous studies had suggested that selenoneine - a novel selenocompound - may be accumulating
Francisco Barahona et al.
Analytical chemistry, 87(5), 3039-3047 (2015-01-30)
This work proposes the use of multimodal mixtures of monodispersed silica nanoparticles (SiO2-NPs) standards for the simultaneous determination of size and concentration of SiO2-NPs in aqueous suspensions by asymmetric flow field-flow fractionation (AF4) coupled to inductively coupled plasma mass spectrometry
Heat of absorption of CO2 in aqueous ammonia and ammonium carbonate/carbamate solutions.
Qin F, et al.
International Journal of Greenhouse Gas Control, 5(3), 405-412 (2011)
N N Mal'tseva et al.
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993), 62(3), 56-65 (2000-08-10)
The intensification of activity of associative and symbiotic nitrogen fixation while using low amounts of ammonium-carbonate compounds, carbonic-ammonium salts, in particular, has been established. The main ways and mechanisms of the action of ammonium-carbonate salts as a new and perspective

Protocols

Protocol for LC/MS Analysis of Glyphosate and Metabolites on apHera™ NH2, 2 mm I.D. Column

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service