Skip to Content
Merck
All Photos(2)

Documents

P0483

Sigma-Aldrich

Anti-Potassium Channel SK2 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Apamin-Sensitive Conductance Ca2+-Dependent K+ Channel, KCa2, Kcnn2, SKCa2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human, rat

technique(s)

western blot (chemiluminescent): 1:200 using rat brain membranes

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Potassium calcium-activated channel subfamily N member 2 (KCNN2) or SK2 is expressed in rat Purkinje cells. The gene encoding this protein is localized on rat chromosome 18q11.

Immunogen

synthetic peptide corresponding to amino acids 542-559 of rat potassium channel SK2.

Application

Anti-Potassium Channel SK2 (Apamin-Sensitive Conductance Ca2+-Dependent K+ Channel, KCa2, Kcnn2, SKCa2) antibody produced in rabbit has been used for immunohistochemistry and immunofluorescence.

Biochem/physiol Actions

Potassium calcium-activated channel subfamily N member 2 (KCNN2) or SK2 has a role in the modulation of intrinsic excitability of neurons and Purkinje cells. It also has a role in synaptic transmission and plasticity. SK2 promotes hippocampus-dependent learning and memory in the mammalian brain.

Physical form

Lyophilized at 0.3-1.4 mg/mL from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.025% - 0.050% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Low-Level Vagus Nerve Stimulation Upregulates Small Conductance Calcium Activated Potassium Channels in the Stellate Ganglion
Mark J. Shen
Heart Rhythm : the Official Journal of the Heart Rhythm Society (2013)
SK2 channel expression and function in cerebellar Purkinje cells
Eric Hosy
The Journal of Physiology (2011)
Neurotransmitter modulation of small conductance Ca2+-activated K+ (SK) channels by regulation of Ca2+ gating
Francois M
Neuron (2008)
Eleonora Katz et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 24(36), 7814-7820 (2004-09-10)
In the mature cochlea, inner hair cells (IHCs) transduce acoustic signals into receptor potentials, communicating to the brain by synaptic contacts with afferent fibers. Before the onset of hearing, a transient efferent innervation is found on IHCs, mediated by a
Jing-Yi Jeng et al.
The Journal of physiology, 598(18), 3891-3910 (2020-07-02)
Age-related hearing loss (ARHL) is a very heterogeneous disease, resulting from cellular senescence, genetic predisposition and environmental factors (e.g. noise exposure). Currently, we know very little about age-related changes occurring in the auditory sensory cells, including those associated with the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service