Skip to Content
Merck
All Photos(1)

Documents

T7408

Sigma-Aldrich

Triethylammonium bicarbonate buffer

1.0 M, pH 8.5±0.1

Synonym(s):

Triethylammonium hydrogen carbonate buffer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
3624751
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

form

liquid

storage condition

(Tightly closed)

concentration

1.0 M

color

colorless

pH

8.5±0.1

density

1.02 g/mL at 25 °C (lit.)

suitability

suitable for LC-MS
suitable for mass spectrometry

application(s)

diagnostic assay manufacturing
microbiology
sample preparation

storage temp.

2-8°C

SMILES string

OC(O)=O.CCN(CC)CC

InChI

1S/C6H15N.CH2O3/c1-4-7(5-2)6-3;2-1(3)4/h4-6H2,1-3H3;(H2,2,3,4)

InChI key

AFQIYTIJXGTIEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triethylammonium bicarbonate buffer is used in ion-exchange chromatography and electrophoresis.
It has also been used:
  • in trypsin digestion
  • as a component of lysis buffer
  • to adjust the pH of buffer for protein quantification

Buffer for use in ion-exchange chromatography and electrophoresis.

Biochem/physiol Actions

Triethylammonium bicarbonate buffer is useful in oligonucleotides purification for electrospray mass spectrometry analysis. It is also utilized in trypsin digestion buffers.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

comparable product

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of Rosmarinic Acid-Adducted Sites in Meat Proteins in a Gel Model under Oxidative Stress by Triple TOF MS/MS.
Tang CB, et al.
Journal of Agricultural and Food Chemistry, 64(33), 6466-6476 (2016)
Differential protein expression analysis of degenerative aortic stenosis by iTRAQ labeling.
Alonso-Orgaz S, et al.
Methods in Molecular Biology, 109-117 (2013)
Stefani N Thomas et al.
iScience, 23(6), 101079-101079 (2020-06-14)
The National Cancer Institute (NCI) Clinical Proteomic Tumor Analysis Consortium (CPTAC) established a harmonized method for large-scale clinical proteomic studies. SWATH-MS, an instance of data-independent acquisition (DIA) proteomic methods, is an alternate proteomic approach. In this study, we used SWATH-MS
Marco L Hennrich et al.
Analytical chemistry, 81(18), 7814-7822 (2009-08-20)
In proteomics, proteolytic peptides are often chemically modified to improve MS analysis, peptide identification, and/or to enable protein/peptide quantification. It is known that such chemical modifications can alter peptide fragmentation in collision induced dissociation MS/MS. Here, we investigated the fragmentation
Evaluation of the Effect of Trypsin Digestion Buffers on Artificial Deamidation
Hao P, et al.
Journal of Proteome Research, 14(2), 1308-1314 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service