Skip to Content
Merck
All Photos(1)

Key Documents

90360

Sigma-Aldrich

Triethylammonium bicarbonate buffer

volatile buffer, ~1.0 M in H2O

Synonym(s):

Buffer solution 1 M pH 8.5 (volatile), Triethylammonium hydrogen carbonate buffer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
3624751
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Quality Level

form

liquid

concentration

~1.0 M in H2O

refractive index

n20/D 1.355

pH

8.4-8.6 (25 °C)

density

1.018 g/mL at 20 °C

λ

neat

UV absorption

λ: 260 nm Amax: ≤0.05
λ: 280 nm Amax: ≤0.05

application(s)

diagnostic assay manufacturing

storage temp.

2-8°C

SMILES string

OC(O)=O.CCN(CC)CC

InChI

1S/C6H15N.CH2O3/c1-4-7(5-2)6-3;2-1(3)4/h4-6H2,1-3H3;(H2,2,3,4)

InChI key

AFQIYTIJXGTIEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Buffer for use in ion-exchange chromatography and electrophoresis.

Other Notes

For use in ion-exchange chromatography and electrophoresiscitation
For use in ion-exchange chromatography and electrophoresis

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dwayne A Elias et al.
Nucleic acids research, 37(9), 2926-2939 (2009-03-19)
Hypothetical (HyP) and conserved HyP genes account for >30% of sequenced bacterial genomes. For the sulfate-reducing bacterium Desulfovibrio vulgaris Hildenborough, 347 of the 3634 genes were annotated as conserved HyP (9.5%) along with 887 HyP genes (24.4%). Given the large
Marco L Hennrich et al.
Analytical chemistry, 81(18), 7814-7822 (2009-08-20)
In proteomics, proteolytic peptides are often chemically modified to improve MS analysis, peptide identification, and/or to enable protein/peptide quantification. It is known that such chemical modifications can alter peptide fragmentation in collision induced dissociation MS/MS. Here, we investigated the fragmentation
C W Mahoney et al.
Analytical biochemistry, 138(1), 246-251 (1984-04-01)
The separation of 5'-adenosine di- and triphosphates from inorganic pyrophosphate or imidodiphosphate can be accomplished with reverse-phase HPLC by using a solvent system buffered by triethylammonium bicarbonate (pH 6.7). This buffer was used because it was neutral, readily volatile at
Use of triethlammonium buffers in ion-exchange chromatography and electrophoresis.
J PORATH
Nature, 175(4454), 478-478 (1955-03-12)
D Lemaire et al.
Analytical chemistry, 73(8), 1699-1706 (2001-05-08)
The use of triethylammonium bicarbonate (TEAB) solution in electrospray mass spectrometry proved to be a very efficient way for studying proteins or noncovalent protein complexes under "nondenaturing" conditions. The low charge states observed in the mass spectra improve the separation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service