Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352204
NACRES:
NA.78
Form:
ready-to-use solution
Recombinant:
expressed in Pichia pastoris
Product Name
SOLu-Trypsin, recombinant, expressed in Pichia pastoris, Proteomics Grade, liquid
recombinant
expressed in Pichia pastoris
Quality Segment
grade
Proteomics Grade
description
Activity:≥ 10,000 unit/mg protein
form
ready-to-use solution
suitability
suitable for mass spectrometry
UniProt accession no.
shipped in
wet ice
storage temp.
2-8°C
General description
Trypsin is a pancreatic serine endoprotease. SOLu-Trypsin is made from recombinant trypsin, porcine sequence. It does not possess chymotryptic activity.
Application
SOLu-Trypsin:
- has been used to digest the gel pieces for mass spectrometry identification of active deubiquitnases (DUBs)
- has been used in trypsin sensitivity assay
- may be used for the digestion of wine proteins
Biochem/physiol Actions
- SOLu-Trypsin (EMS0004) is our exclusive, Advanced Proteomics Grade enzyme that is solution-stable for mass spectrometry.
- Designed to be stable in solution when refrigerated, SOLu-Trypsin can be used immediately without preparation.
- Other forms of trypsin require thawing or reconstitution, and must be discarded if not used immediately.
- SOLu-Trypsin allows excess product to be saved for future use, thus eliminating unnecessary waste and cost.
- It is formulated with a high-purity recombinant trypsin, free of chymotryptic activity, to ensure high fidelity digestion.
Trypsin is used in proteomics research for peptide mapping and protein sequence work due to its highly specific cleavage resulting in a limited number of tryptic peptides. It hydrolyzes peptide bonds specifically at the carboxyl side of arginine and lysine residues.
Features and Benefits
- Ready to use - no preparation, such as reconstitution or thawing, is required
- Fits seamlessly into established workflow - no need to modify protocols
- Eliminates waste - remains stable in the refrigerator after use so there is no need to discard excess product
- Recombinant, porcine sequence - no chymotryptic activity
- Stable for short-term use at room temperature in an autosampler or on a liquid handling robot
Still not finding the right product?
Explore all of our products under SOLu-Trypsin
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Global Trade Item Number
| SKU | GTIN |
|---|---|
| EMS0004-4X100UG | 04061822415751 |
| EMS0004-100UG | 04061833253205 |