Skip to Content
Merck
All Photos(3)

Key Documents

M6545

Sigma-Aldrich

Mitoxantrone dihydrochloride

≥97% (HPLC)

Synonym(s):

1,4-Dihydroxy-5,8-bis[[2-[(2-hydroxyethyl)amino]ethyl]amino]-9,10-anthracenedione dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28N4O6 · 2HCl
CAS Number:
Molecular Weight:
517.40
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.25

Assay

≥97% (HPLC)

SMILES string

Cl[H].Cl[H].OCCNCCNc1ccc(NCCNCCO)c2C(=O)c3c(O)ccc(O)c3C(=O)c12

InChI

1S/C22H28N4O6.2ClH/c27-11-9-23-5-7-25-13-1-2-14(26-8-6-24-10-12-28)18-17(13)21(31)19-15(29)3-4-16(30)20(19)22(18)32;;/h1-4,23-30H,5-12H2;2*1H

InChI key

ZAHQPTJLOCWVPG-UHFFFAOYSA-N

Gene Information

human ... TOP2A(7153)

Looking for similar products? Visit Product Comparison Guide

Application

Mitoxantrone dihydrochloride has been used:
  • to induce calreticulin surface expression in cells
  • as a genotoxin agent
  • as a topoisomerase inhibitor

Biochem/physiol Actions

Mitoxantrone is a cytostatic anthracenedione that intercalates in DNA and increases the incidence of double-strand breaks by stabilizing the cleavable complex of topoisomerase II and DNA. Mitoxantrone also displays broad immunosuppressive activity inhibiting proliferation of all classes of lymphocytes and inducing apoptosis of antigen-presenting T cells. It used clinically as a chemotherapeutic agent against leukemias and solid tumors and as an immune system modulator in multiple sclerosis.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuting Ma et al.
The Journal of experimental medicine, 208(3), 491-503 (2011-03-09)
By triggering immunogenic cell death, some anticancer compounds, including anthracyclines and oxaliplatin, elicit tumor-specific, interferon-γ-producing CD8(+) αβ T lymphocytes (Tc1 CTLs) that are pivotal for an optimal therapeutic outcome. Here, we demonstrate that chemotherapy induces a rapid and prominent invasion
Chyuan-Chuan Wu et al.
Nucleic acids research, 41(22), 10630-10640 (2013-09-17)
Type II topoisomerases (Top2s) alter DNA topology via the formation of an enzyme-DNA adduct termed cleavage complex, which harbors a transient double-strand break in one DNA to allow the passage of another. Agents targeting human Top2s are clinically active anticancer
Adi Tzur-Balter et al.
Acta biomaterialia, 9(4), 6208-6217 (2013-01-01)
Nanostructured porous silicon (PSi) thin films, fabricated by the electrochemical anodization of single crystalline Si wafers, are studied as delivery systems for the anticancer drug mitoxantrone dihydrochloride (MTX). The surface chemistry of the PSi carriers was tailored by surface alkylation
A high through-put screen for small molecules modulating MCM2 phosphorylation identifies Ryuvidine as an inducer of the DNA damage response.
FitzGerald J
PLoS ONE, 9, e98891-e98891 (2014)
James M Stankiewicz et al.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics, 10(1), 77-88 (2012-12-29)
Immunosuppressives have been used in multiple sclerosis (MS) since 1966. Today, we have many treatments for the relapsing forms of the disease, including 8 US Food and Drug Administration-approved therapies, with more soon to be introduced. Given the current treatment

Articles

DNA damage and repair mechanism is vital for maintaining DNA integrity. Damage to cellular DNA is involved in mutagenesis, the development of cancer among others.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service