Skip to Content
Merck
All Photos(4)

Key Documents

F6627

Sigma-Aldrich

5-Fluorouracil

≥99% purity (HPLC), powder

Synonym(s):

2,4-Dihydroxy-5-fluoropyrimidine, 5-FU, 5-Fluoro-2,4(1H,3H)-pyrimidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3FN2O2
CAS Number:
Molecular Weight:
130.08
Beilstein:
127172
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

5-Fluorouracil, ≥99% (HPLC), powder

Quality Level

Assay

≥99% (HPLC)

form

powder

technique(s)

titration: suitable

color

white

mp

282-286 °C (dec.) (lit.)

solubility

1 M NH4OH: soluble
DMSO/DMF: soluble
methanol: soluble

εmax

7.07 at 265 nm in 0.1 M HCl

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

FC1=CNC(=O)NC1=O

InChI

1S/C4H3FN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)

InChI key

GHASVSINZRGABV-UHFFFAOYSA-N

Gene Information

human ... TYMS(7298)

Looking for similar products? Visit Product Comparison Guide

Application

5-Fluorouracil has been used to induce apoptosis in cells. It has been used as a chemosensitizing agent.

Biochem/physiol Actions

A potent antitumor agent that affects pyrimidine synthesis by inhibiting thymidylate synthetase, thus depleting intracellular dTTP pools. It is metabolized to ribonucleotides and deoxyribonucleotides, which can be incorporated into RNA and DNA. Treatment of cells with 5-FU leads to an accumulation of cells in S-phase and has been shown to induce p53 dependent apoptosis.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Drug Nutrient Interactions Nutrition and Health (2004)
Curcumin promotes apoptosis, increases chemosensitivity, and inhibits nuclear factor kappaB in esophageal adenocarcinoma.
Hartojo W
Translational Oncology, 3, 99-99 (2010)
Overexpression of microRNA-125b sensitizes human hepatocellular carcinoma cells to 5-fluorouracil through inhibition of glycolysis by targeting hexokinase II.
Jiang JX
Molecular Medicine Reports, 10, 995-995 (2014)
Coulson CJ
Molecular Mechanisms Of Drug Action (1994)
CCL2/CCR2 chemokine signaling coordinates survival and motility of breast cancer cells through Smad3 protein- and p42/44 mitogen-activated protein kinase (MAPK)-dependent mechanisms.
Fang WB et al.
The Journal of Biological Chemistry, 287, 36593-36593 (2012)

Articles

We presents an article on Autophagy in Cancer Promotes Therapeutic Resistance

DNA damage and repair mechanism is vital for maintaining DNA integrity. Damage to cellular DNA is involved in mutagenesis, the development of cancer among others.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service