Skip to Content
Merck
All Photos(3)

Key Documents

T2577

Sigma-Aldrich

Temozolomide

≥98% (HPLC), powder, DNA methylating agent

Synonym(s):

3,4-Dihydro-3-methyl-4-oxoimidazo[5,1-d]-1,2,3,5-tetrazine-8-carboxamide, 3-Methyl-4-oxo-8-imidazolo[5,1-d][1,2,3,5]tetrazinecarboxamide, 4-Methyl-5-oxo-2,3,4,6,8-pentazabicyclo[4.3.0]nona-2,7,9-triene-9-carboxamide, 8-Carbamoyl-3-methylimidazo[5,1-d]-1,2,3,5-tetrazin-4(3H)-one, NSC 362856

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N6O2
CAS Number:
Molecular Weight:
194.15
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Temozolomide, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 mg/mL, clear
H2O: insoluble

originator

Schering Plough

storage temp.

2-8°C

SMILES string

CN1N=Nc2c(ncn2C1=O)C(N)=O

InChI

1S/C6H6N6O2/c1-11-6(14)12-2-8-3(4(7)13)5(12)9-10-11/h2H,1H3,(H2,7,13)

InChI key

BPEGJWRSRHCHSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Temozolomide is a small lipophilic alkylating agent.[1] It interacts covalently with the nucleophile microenvironment within DNA (Guanine residues). Due to this property temozolomide is considered a cytotoxic agent. The mechanism of action of TMZ is dependent upon its ability to hydrolyze DNA, resulting in DNA degradation and destruction of tumor cells. Malignant Glioma cells respond primarily to TMZ by G2/ M cell cycle arrest or in rare cases by apoptosis.[1] Temozolomide is a DNA methylating agent and drug resistance-modifying agent; anti-tumor and anti-angiogenic. Temozolomide induces G2/M arrest and apoptosis through adduction of a methyl group to O6 position of guanine in genomic DNA and functional inactivation of DNA repair protein O(6)-alkylguanine DNA alkyltransferase (AGT) in base excision repair (BER) pathway.

Application

Temozolomide has been used for analyzing drug resistance mechanisms in glioblastoma cell lines. Temozolomide has been used to induce cytotoxic effects on glioblastoma cells to study the outcome of protein disulfide isomerase (PDI) inhibition.[2]

Biochem/physiol Actions

Temozolomide is a DNA methylating agent and drug resistance-modifying agent; anti-tumor and anti-angiogenic. Temozolomide induces G2/M arrest and apoptosis through adduction of a methyl group to O6 position of guanine in genomic DNA and functional inactivation of DNA repair protein O(6)-alkylguanine DNA alkyltransferase (AGT) in base excision repair (BER) pathway.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Schering Plough. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Temozolomide is soluble in DMSO at a concentration that is greater than 20 mg/ml. It is insoluble in water.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonin Dréan et al.
Journal of neuro-oncology, 138(3), 479-486 (2018-03-10)
ATP-binding cassette transporters (ABC transporters) regulate traffic of multiple compounds, including chemotherapeutic agents, through biological membranes. They are expressed by multiple cell types and have been implicated in the drug resistance of some cancer cells. Despite significant research in ABC
Michael Weller et al.
International journal of cancer, 134(10), 2437-2447 (2014-03-13)
The epidermal growth factor receptor vIII mutant (EGFRvIII) is found in ~50% of all EGFR-amplified glioblastomas and constitutes a tumor-specific therapeutic target. To assess molecular testing approaches and the prognostic role of EGFRvIII in patients treated according to current standards
Leon D Ortiz et al.
Clinics (Sao Paulo, Brazil), 67 Suppl 1, 119-123 (2012-05-25)
Temozolomide is an alkylating agent used in the treatment of gliomas and, more recently, aggressive pituitary adenomas and carcinomas. Temozolomide methylates DNA and, thereby, has antitumor effects. O6-methylguanine-DNA methyltransferase, a DNA repair enzyme, removes the alkylating adducts that are induced
Mark R Gilbert et al.
The New England journal of medicine, 370(8), 699-708 (2014-02-21)
Concurrent treatment with temozolomide and radiotherapy followed by maintenance temozolomide is the standard of care for patients with newly diagnosed glioblastoma. Bevacizumab, a humanized monoclonal antibody against vascular endothelial growth factor A, is currently approved for recurrent glioblastoma. Whether the
T C Hirst et al.
British journal of cancer, 108(1), 64-71 (2013-01-17)
Malignant glioma is an aggressive tumour commonly associated with a dismal outcome despite optimal surgical and radio-chemotherapy. Since 2005 temozolomide has been established as first-line chemotherapy. We investigate the role of in vivo glioma models in predicting clinical efficacy. We

Articles

We presents an article on Autophagy in Cancer Promotes Therapeutic Resistance

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

See All

Questions

1–6 of 6 Questions  
  1. Is undissolved Temozolomide still stable if it was stored at RT for 3 days upon arrival?

    1 answer
    1. This product is stable at ambient temperatures for relatively short periods of time. The item is shipped at ambient temperature. A transit time of up to 2 weeks or a brief temperature excursion will not impact the quality of this material. Upon delivery, this product should be stored at refrigerated at 2 - 8°C.

      Helpful?

  2. How long is this stable for between 2-8 degrees celsius?

    1 answer
    1. This product is not assigned an expiration date. Products that are robust in nature and extremely stable, in the original unopened container, are not assigned an expiration date or retest schedule. These products will have no dating reported on either the label or the Certificate of Analysis.
      Products that are not assigned an expiration or retest date are covered by a warranty period of 1 year from the date of product shipment.
      Full Product Dating Information may be reviewed at the link below: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/190/408/product-dating-information-ms.pdf

      Helpful?

  3. How long is Temozolomide stable for when dissolved as recommended, in DMSO at 10 mg/mL and storaged at 2-8C? Also, can the solution only be stored at 2-8C or can it be stored at -20C or -80C for long term storage

    1 answer
    1. Wherever possible, solutions should be prepared fresh and used on the same day. However, should stock solutions need to be prepared in advance, store in aliquots at -20°C for up to 1 month. Allow aliquots to equilibrate to room temperature prior to opening. The long term storage at other temperatures is not determined. However, the published reference below may prove helpful:
      Link: https://onlinelibrary.wiley.com/doi/abs/10.1002/j.2055-2335.2012.tb00145.x

      Helpful?

  4. Do you have a reference for the analysis of Temozolomide (Product No. T2577) in serum?

    1 answer
    1. We have not confirmed the method in our labs here at Sigma, but see: Kim, H. et al., High-performance liquid chromatographic analysis and stability of anti-tumor agent temozolomide in human plasma. Journal of Pharmaceutical and Biomedical Analysis, 24(3), 461-468 (2001).

      Helpful?

  5. Is Temozolomide (Product No. T2577) an active drug or a prodrug?

    1 answer
    1. Temozolomide is considered a prodrug. It is hydrolyzed in vivo to MTIC (monomethyl triazeno imidazole carboxamide). See: Kim, H. et al., High-performance liquid chromatographic analysis and stability of anti-tumor agent temozolomide in human plasma. Journal of Pharmaceutical and Biomedical Analysis, 24(3), 461-468 (2001).

      Helpful?

  6. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service