Skip to Content
Merck
All Photos(1)

Key Documents

M4892

SAFC

McCoy′s 5A Medium

Modified, with L-glutamine, without sodium bicarbonate, powder, suitable for cell culture

Pharma Manufacturing

Synonym(s):

5A basal medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71
Pricing and availability is not currently available.

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

components

phenol red: 0.011 g/L
L-glutamine: 0.21915 g/L
glucose: 3.0 g/L (Dextro)
NaHCO3: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

McCoy′s 5A Medium is recommended to produce viruses in primary cell cultures.

Application

McCoy′s 5A Medium has been used for culturing:
  • human colon carcinoma cell lines (HCT116, RKO, FET, CBS, HCT116b, and TENN) to study the function of leucine-rich repeat-containing G-protein-coupled receptor (LGR5) in colon cancer metastasis
  • human colorectal HCT116 cells to study miR-224 over-expression in cancer
  • osteoblasts to study ion reactivity of calcium-deficient hydroxyapatite in standard cell culture media

Biochem/physiol Actions

McCoy′s 5A Medium has been originally developed to support liver tumor cells by modification of the amino acids found in BME. This formulation has also been used to support the growth of primary cultures of bone marrow, skin, gingiva, kidney, omentum, adrenals, lung, spleen, rat embryos, and other cell types.

Quantity

Formulated to contain 11.9 grams of powder per liter of medium.

Reconstitution

Supplement with 2.2 g/L sodium bicarbonate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Liu et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 44(4), 1396-1410 (2017-12-05)
Osteosarcoma (OS) is the most common primary malignant tumor of bone tissue. Although treatment effectiveness has improved, the OS survival rate has fluctuated in recent years. Andrographolide (AG) has been reported to have antitumor activity against a variety of tumors.
Yan Zhang et al.
Oncotarget, 8(53), 91134-91149 (2017-12-07)
Human osteosarcoma is considered a malignant tumor with poor prognosis that readily metastasizes. Tetrahydrocurcumin (THC) has been reported to have anti-tumor activity in numerous tumors. In addition, hypoxia-inducible factor-1α (HIF-1α) has been demonstrated to be associated with tumor metastasis by
Yu Wang et al.
PloS one, 8(7), e68744-e68744 (2013-08-08)
MicroRNA-224 (miR-224) is frequently over-expressed in liver and colorectal cancers. We and others have previously described the role of miR-224 over-expression in cell proliferation in vitro but we have yet to identify the relevant miR-224 direct target. In this study
Archana Kumari et al.
The EMBO journal, 39(14), e104006-e104006 (2020-06-23)
Cellular studies of filamentous actin (F-actin) processes commonly utilize fluorescent versions of toxins, peptides, and proteins that bind actin. While the choice of these markers has been largely based on availability and ease, there is a severe dearth of structural
Xin Zhang et al.
Cellular and molecular gastroenterology and hepatology, 12(4), 1179-1199 (2021-06-05)
Chronic inflammation in colon section is associated with an increased risk of colorectal cancer (CRC). Proinflammatory cytokines were produced in a tumor microenvironment and correlated with poor clinical outcome. Tumor-infiltrating T cells were reported to be greatly involved in the

Articles

McCoy's 5A Modified Media Formulation

Protocols

Powdered media and salt mixtures are extremely hygroscopic and should be protected from atmospheric moisture.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service