Skip to Content
Merck
All Photos(1)

Documents

L1518

Sigma-Aldrich

L-15 Medium (Leibovitz)

With L-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Leibovitz L15

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

phenol red: 0.011 g/L
L-glutamine: 0.3 g/L
sodium pyruvate: 0.55 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

L-15 Medium (Leibovitz) has been originally formulated for use in carbon dioxide-free systems requiring sodium bicarbonate supplements. L-15 supports established cell lines as well as primary explants of embryonic and adult human tissue.

Application

L-15 Medium (Leibovitz) has been used to culture WM35 cell lines and the gill cell line, RTgill-W1.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Remi L Gratacap et al.
BMC biotechnology, 20(1), 35-35 (2020-06-25)
Genome editing is transforming bioscience research, but its application to non-model organisms, such as farmed animal species, requires optimisation. Salmonids are the most important aquaculture species by value, and improving genetic resistance to infectious disease is a major goal. However
Jithine Jayakumar Rajeswari et al.
Scientific reports, 10(1), 6264-6264 (2020-04-15)
Phoenixin-20 (PNX-20) is a bioactive peptide with hormone-like actions in vertebrates. In mammals, PNX stimulates hypothalamo-pituitary-gonadal hormones and regulate reproductive processes. Our immunohisto/cytochemical studies show PNX-like and the putative PNX receptor, SREB3-like immunoreactivity in the gonads of zebrafish, and in
Screening of Chilean fish-killing microalgae using a gill cell-based assay
Mardones JI
Latin American Journal of Aquatic Research, 48, 329-335 (2020)
Charlotte Falaise et al.
Aquatic toxicology (Amsterdam, Netherlands), 209, 13-25 (2019-01-27)
Marennine is a water-soluble blue-green pigment produced by the marine diatom Haslea ostrearia. The diatom and its pigment are well known from oyster farming areas as the source of the greening of oyster gills, a natural process increasing their market
Fish gill damage by the dinoflagellate Alexandrium catenella from Chilean fjords: Synergistic action of ROS and PUFA
Mardones JI, et al.
Harmful Algae, 49, 40-49 (2015)

Articles

L-15 Media Formulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service