Skip to Content
Merck
All Photos(3)

Key Documents

398152

Sigma-Aldrich

Lithium sulfate monohydrate

ACS reagent, ≥99.0% dry basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2SO4 · H2O
CAS Number:
Molecular Weight:
127.96
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥99.0% dry basis
grade:
ACS reagent
form:
powder or crystals
solubility:
water: soluble 128 g/L at 20 °C

grade

ACS reagent

Quality Level

Assay

≥99.0% dry basis

form

powder or crystals

impurities

≤0.01% insolubles

loss

13.0-15.0% loss on drying

pH

5-7 (25 °C, 128 g/L)

solubility

water: soluble 128 g/L at 20 °C

anion traces

chloride (Cl-): ≤0.002%
nitrate (NO3-): ≤0.001%

cation traces

Fe: ≤0.001%
K: ≤0.05%
Na: ≤0.05%
heavy metals: ≤0.001% (by ICP)

SMILES string

[Li+].[Li+].O.[O-]S([O-])(=O)=O

InChI

1S/2Li.H2O4S.H2O/c;;1-5(2,3)4;/h;;(H2,1,2,3,4);1H2/q2*+1;;/p-2

InChI key

RKGLUDFWIKNKMX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Lithium sulfate monohydrate is an inorganic lithium salt. It is relatively stable in atmospheric conditions. It is used in building blocks and chemical synthesis.

Application

Lithium sulfate monohydrate is used to study the interference on the salicylate method, which is used for quantifying ammonia.
Lithium sulfate monohydrate may be used in the preparation of Cu(II) doped lithium sulfate monohydrate (LSMH) crystal.

Features and Benefits

Lithium sulfate monohydrate has the highest pyroelectric coefficient and remarkable piezoelectric and electro-optic properties.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neutron diffraction structural study of pyroelectric Li2SO4?H2O at 293, 80, and 20K.
Lundgren JO, et al.
J. Chem. Phys. , 80(1), 423-430 (1984)
Nonlinear optical properties of lithium sulfate monohydrate.
Bayarjargal L.
Crystal Research and Technology, 43(11), 1138-1142 (2008)
Growth and characterization of Cu (II) doped negatively soluble lithium sulfate monohydrate crystals.
Boopathi K, et al.
Journal of Crystal Growth, 386, 32-37 (2014)
Pyroelectric coefficient of lithium sulfate monohydrate (4.2-320 K).
Lang SB.
Physical Review. B, Condensed Matter and Materials Physics, 4(10), 3603-3603 (1971)
Eleonora Turco et al.
Molecular cell, 74(2), 330-346 (2019-03-12)
The autophagy cargo receptor p62 facilitates the condensation of misfolded, ubiquitin-positive proteins and their degradation by autophagy, but the molecular mechanism of p62 signaling to the core autophagy machinery is unclear. Here, we show that disordered residues 326-380 of p62

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service