Skip to Content
Merck
All Photos(2)

Key Documents

203653

Sigma-Aldrich

Lithium sulfate

greener alternative

≥99.99% trace metals basis

Synonym(s):

Dilithium sulfate, Lithiophor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2SO4
CAS Number:
Molecular Weight:
109.94
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38030404
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.99% trace metals basis
form:
solid

Quality Level

Assay

≥99.99% trace metals basis

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤100.0 ppm Trace Metal Analysis

mp

845 °C (lit.)

density

2.22 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li+].[Li+].[O-]S([O-])(=O)=O

InChI

1S/2Li.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

INHCSSUBVCNVSK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Lithium sulfate can be used:
  • In the pretreatment of graphite electrodes of lithium-ion batteries(LIBs). LiSO4-treated electrodes show excellent cycling performance and low interfacial impedance resulting in faster charging of LIBs.
  • As a precursor to synthesize sheet-like Li2S@C nanocomposites which are cathode materials for LIBs.
  • As an electrolyte for intermediate temperature fuel cells.
  • As a processing additive to protect Ni-rich layered oxide cathodes, resulting in improved electrochemical performance.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pretreatment of Graphite Anodes with Lithium Sulfate to Improve the Cycle Performance of Lithium-Ion Batteries
Xiaoling Cui, et al.
Energy Technology, 5, 549-556 (2017)
Structure and ionic conductivity of lithium sulphate-aluminum oxide ceramics
B. Zhu, et al.},
Solid State Ionics, 70-71, 125-129 (1994)
A high energy density Li2S@ C nanocomposite cathode with a nitrogen-doped carbon nanotube top current collector
Su Zhang, et al.
Journal of Material Chemistry A, 3, 18913-18919 (2015)
Artur T Cordeiro et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 62(Pt 10), 1049-1051 (2006-10-03)
Dihydroorotate dehydrogenases (DHODHs) are flavin-containing enzymes that catalyze the oxidation of L-dihydroorotate to orotate, the fourth step in the de novo pyrimidine nucleotide synthesis pathway. In this study, DHODH from Leishmania major has been crystallized by the vapour-diffusion technique using
Dinesh Rangappa et al.
Nano letters, 12(3), 1146-1151 (2012-02-16)
Novel ultrathin Li(2)MnSiO(4) nanosheets have been prepared in a rapid one pot supercritical fluid synthesis method. Nanosheets structured cathode material exhibits a discharge capacity of ~340 mAh/g at 45 ± 5 °C. This result shows two lithium extraction/insertion performances with

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service