Skip to Content
Merck
All Photos(1)

Key Documents

62613

Sigma-Aldrich

Lithium sulfate

purum p.a., ≥98.0% (T)

Synonym(s):

Dilithium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2SO4
CAS Number:
Molecular Weight:
109.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

purum p.a.

Quality Level

Assay

≥98.0% (T)

form

powder

loss

≤2% loss on drying, 200 °C

mp

845 °C (lit.)

density

2.22 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

Ca: ≤100 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤200 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Li+].[Li+].[O-]S([O-])(=O)=O

InChI

1S/2Li.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

INHCSSUBVCNVSK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Artur T Cordeiro et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 62(Pt 10), 1049-1051 (2006-10-03)
Dihydroorotate dehydrogenases (DHODHs) are flavin-containing enzymes that catalyze the oxidation of L-dihydroorotate to orotate, the fourth step in the de novo pyrimidine nucleotide synthesis pathway. In this study, DHODH from Leishmania major has been crystallized by the vapour-diffusion technique using
Jun-ichi Edahiro et al.
Journal of bioscience and bioengineering, 100(4), 449-454 (2005-11-29)
A rapid and simple selection method of high-yield cells has been desired to establish highly productive cell lines for useful secondary metabolites. For this purpose, a new attempt was made to partition cultured plant cells in a poly(ethylene glycol)-dextran aqueous
Dinesh Rangappa et al.
Nano letters, 12(3), 1146-1151 (2012-02-16)
Novel ultrathin Li(2)MnSiO(4) nanosheets have been prepared in a rapid one pot supercritical fluid synthesis method. Nanosheets structured cathode material exhibits a discharge capacity of ~340 mAh/g at 45 ± 5 °C. This result shows two lithium extraction/insertion performances with
Benedikt Habermeyer et al.
Psychiatrische Praxis, 35(4), 198-200 (2007-11-08)
Lithium is known to be neurotoxic at higher serum levels. In rare cases patients develop symptoms of intoxication or full blown intoxications even when lithium levels are normal. Our aim was to detect predicting factors of lithium neurotoxicity at normal
L K Singh et al.
The National medical journal of India, 24(3), 151-152 (2011-07-27)
Lithium-induced cardiotoxicity, though rare at therapeutic levels, has been reported frequently in overdoses. We report a patient who developed sinus bradycardia while being treated with lithium carbonate even though the serum lithium levels were within the therapeutic range. It reversed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service