Skip to Content
Merck
All Photos(4)

Key Documents

326755

Sigma-Aldrich

Copper(II) acetate

greener alternative

98%

Synonym(s):

Cupric acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(CO2CH3)2
CAS Number:
Molecular Weight:
181.63
Beilstein:
3595638
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
form:
powder or crystals

vapor density

6.9 (vs air)

Quality Level

Assay

98%

form

powder or crystals

reaction suitability

reaction type: click chemistry

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

CC(=O)O[Cu]OC(C)=O

InChI

1S/2C2H4O2.Cu/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

OPQARKPSCNTWTJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Copper(II) acetate also known as cupric acetate, can be used as a catalyst in various processes in the field of greener chemistry. It is particularly useful in cross-coupling reactions, where it can promote the formation of carbon-carbon or carbon-heteroatom bonds, without the need for hazardous reagents or solvents

Application

Catalyst for greener amine synthesis by reductive amination with hydrogen gas.

Copper-catalyzed reductive amination of aromatic and aliphatic ketones with anilines using environmental-friendly molecular hydrogen

Copper(II) acetate is used as a catalyst:
  • In the N-arylation of α-amino esters with p-tolylboronic acid to synthesize biaryls via cross-coupling reactions
  • In the the synthesis of substituted isoxazole derivatives

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-Arylation of ?-aminoesters with p-tolylboronic acid promoted by copper (II) acetate
PYS Lam, et al.
Tetrahedron Letters, 44, 1691-1694 (2003)
An efficient synthesis of novel isoxazole bearing pyrazole derivatives via [3+ 2] heteroannulation using cupric acetate
B Audichya, et al.
Journal of Heterocyclic Chemistry, 59, 341-350 (2022)
Anitha M Thomas et al.
Journal of controlled release : official journal of the Controlled Release Society, 150(2), 212-219 (2010-11-26)
5-Fluorouracil (5-FU) is a small, very membrane permeable drug that is poorly retained within the aqueous compartment of liposomal nanoparticles (LNP). To address this problem a novel method relying on formation of a ternary complex comprising copper, low molecular weight
Hanmei Zheng et al.
The Journal of organic chemistry, 74(2), 943-945 (2008-12-11)
A novel copper-catalyzed protocol for the synthesis of carbinol derivatives has been developed. In the presence of copper(II) acetate and dppf, carbinol derivatives were prepared by the addition of arylboronic acids to aromatic aldehydes in good to excellent yields. Moreover
Cinthia da S Lisboa et al.
The Journal of organic chemistry, 76(13), 5264-5273 (2011-05-25)
The oxidative addition of anilines (2) with 1,4-naphthoquinone (3) to give N-aryl-2-amino-1,4-naphthoquinones (1) was found to be catalyzed by copper(II) acetate. In the absence of the catalyst, the reactions are slower and give lower yields with the formation of many

Articles

Copper metal deposition processes are an essential tool for depositing interconnects used in microelectronic applications, giving group 11 (coinage metals: Copper, Silver, and Gold) an important place in atomic layer deposition (ALD) process development.

Explore the principles and applications of click chemistry in drug discovery, highlighting efficient reactions that streamline the synthesis of bioactive compounds.

Explore the principles and applications of click chemistry in drug discovery, highlighting efficient reactions that streamline the synthesis of bioactive compounds.

Explore the principles and applications of click chemistry in drug discovery, highlighting efficient reactions that streamline the synthesis of bioactive compounds.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service