Skip to Content
Merck
All Photos(2)

Key Documents

920339

Sigma-Aldrich

Lithium sulfate

greener alternative

anhydrous, 99.5% trace metals basis

Synonym(s):

″Sulfuric acid, dilithium salt″, Dilithium sulfate, Lithium sulphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2SO4
CAS Number:
Molecular Weight:
109.94
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
grade:
anhydrous
form:
crystals

grade

anhydrous

Quality Level

Assay

99.5% trace metals basis

form

crystals

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

845 °C (lit.)

density

2.22 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[Li+].[Li+].[O-]S([O-])(=O)=O

InChI

1S/2Li.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

INHCSSUBVCNVSK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information

Lithium sulfate is an anhydrous lithium salt with excellent water. It is used as a source of lithium ions in lithium-ion batteries, which are widely used in portable electronic devices and electric vehicles due to their high energy density and long cycle life. In fuel cells, lithium sulfate can be used as an electrolyte material due to its ionic conductivity, which is essential for the efficient operation of the fuel cell.

Application

Lithium sulfate can be:
  • Used as an additive in lead acid batteries. The addition of lithium sulfate improves the cycle life and the efficiency of lead-acid batteries, which are used in various industries, including automotive and energy storage.
  • Used in the fabrication of thin-film solar cells. It can be employed as a dopant to improve the performance of cadmium sulfide (CdS) thin-film solar cells by enhancing their electrical properties.
  • Used in the production of lithium-ion batteries. It is a precursor in the synthesis of lithium iron phosphate (LiFePO4) cathode materials, which are widely used in lithium-ion batteries for their high energy density, long cycle life, and enhanced safety.
  • Used as an electrolyte in the electrochemical characterization of synthesized a-MnO2 for supercapacitor applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia-Yan Luo et al.
Nature chemistry, 2(9), 760-765 (2010-08-24)
Aqueous lithium-ion batteries may solve the safety problem associated with lithium-ion batteries that use highly toxic and flammable organic solvents, and the poor cycling life associated with commercialized aqueous rechargeable batteries such as lead-acid and nickel-metal hydride systems. But all
Ultrathin all-solid-state supercapacitor devices based on chitosan activated carbon electrodes and polymer electrolytes.
Genovese M, et al.
Electrochimica Acta, 273, 392-401 (2018)
Electrochemical Performance of MnO2 Nanorods in Neutral Aqueous Electrolytes as a Cathode for Asymmetric Supercapacitors.
Qu Q, et al.
The Journal of Physical Chemistry C, 113(31), 14020-14027 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service