Skip to Content
Merck
All Photos(1)

Key Documents

1.05697

Supelco

Lithium sulfate monohydrate

99.99 Suprapur®

Synonym(s):

Lithium sulfate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Li2O4S · xH2O
CAS Number:
Molecular Weight:
109.94 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-820-4
NACRES:
NB.21
grade:
for inorganic trace analysis
form:
solid
solubility:
340 g/L

grade

for inorganic trace analysis

Quality Level

Assay

99.99%

form

solid

potency

613 mg/kg LD50, oral (Rat)

pH

4.5-6.0 (20 °C, 50 g/L in H2O)

mp

120 °C

solubility

340 g/L

density

2.05 g/cm3 at 20 °C

bulk density

940 kg/m3

storage temp.

2-30°C

SMILES string

[Li+].[Li+].[S](=O)(=O)([O-])[O-].O

InChI

1S/2Li.H2O4S.H2O/c;;1-5(2,3)4;/h;;(H2,1,2,3,4);1H2/q2*+1;;/p-2

InChI key

RKGLUDFWIKNKMX-UHFFFAOYSA-L

Related Categories

Analysis Note

Purity (metallic): ≥ 99.99 %
Assay (potentiometric): ≥ 99.0 %
Al (Aluminium): ≤ 0.05 ppm
Ba (Barium): ≤ 5 ppm
Ca (Calcium): ≤ 10 ppm
Cd (Cadmium): ≤ 0.010 ppm
Ce (Cerium): ≤ 0.010 ppm
Co (Cobalt): ≤ 0.010 ppm
Cr (Chromium): ≤ 0.010 ppm
Cs (Cesium): ≤ 10 ppm
Cu (Copper): ≤ 0.010 ppm
Eu (Europium): ≤ 0.010 ppm
Fe (Iron): ≤ 0.05 ppm
K (Potassium): ≤ 10 ppm
La (Lanthanum): ≤ 0.010 ppm
Mg (Magnesium): ≤ 1.0 ppm
Mn (Manganese): ≤ 0.05 ppm
Na (Sodium): ≤ 5 ppm
Ni (Nickel): ≤ 0.010 ppm
Pb (Lead): ≤ 0.05 ppm
Rb (Rubidium): ≤ 10 ppm
Sc (Scandium): ≤ 0.010 ppm
Sm (Samarium): ≤ 0.010 ppm
Sr (Strontium): ≤ 5 ppm
Tl (Thallium): ≤ 0.01 ppm
Y (Yttrium): ≤ 0.010 ppm
Yb (Ytterbium): ≤ 0.010 ppm
Zn (Zinc): ≤ 0.20 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service