Skip to Content
Merck
All Photos(1)

Documents

48486

Supelco

Vinyl acetate

analytical standard

Synonym(s):

Acetoxyethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH=CH2
CAS Number:
Molecular Weight:
86.09
Beilstein:
1209327
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Agency

EPA 8240,8260

vapor density

3 (vs air)

vapor pressure

88 mmHg ( 20 °C)

CofA

current certificate can be downloaded

autoignition temp.

801 °F

expl. lim.

13.4 %

packaging

pkg of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.395 (lit.)

bp

72-73 °C (lit.)

mp

−93 °C (lit.)

density

0.934 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care
petroleum

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)OC=C

InChI

1S/C4H6O2/c1-3-6-4(2)5/h3H,1H2,2H3

InChI key

XTXRWKRVRITETP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here
Vinyl acetate is an important ester of vinyl alcohol, which can find applications in the production of homo and copolymer dispersions. The production of vinyl acetate involves the addition of acetic acid to acetylene in the gas phase in the presence of zinc acetate/charcoal as catalyst.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Vinyl acetate may be used as an analytical reference standard for the quantification of the analyte in water samples using liquid chromatography technique.

Other Notes

Contains 3-20 ppm hydroquinone as an inhibitor

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Carc. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

17.6 °F - closed cup

Flash Point(C)

-8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of plastic monomers in water by solid-phase microextraction coupled with liquid chromatography
Batlle R, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 84(2), 431-436 (2001)
Weissermel K and Arpe H-J
Industrial Organic Chemistry (2003)
Anna Piperno et al.
The Journal of organic chemistry, 75(9), 2798-2805 (2010-04-17)
A new template of C-4'-truncated phosphonated nucleosides (TPCOANs) has been obtained in good yields according to two different routes which exploit the reactivity of a phosphonated nitrone. The one-step procedure based on the 1,3-dipolar cycloaddition of a phosphonated nitrone with
Roberto M Risi et al.
Organic letters, 14(4), 1180-1182 (2012-02-07)
A highly atom-economical total synthesis of (+)-patulolide C has been accomplished in three steps from the known (2R)-8-nonyn-2-ol in 49% overall yield and 93% de. A Rh(I)-catalyzed asymmetric hydroformylation (AHF)/ intramolecular Wittig olefination cascade was utilized to set the C4-hydroxyl
B Picquet-Varrault et al.
Environmental science & technology, 44(12), 4615-4621 (2010-05-25)
Vinyl acetate is widely used in industry. It has been classified as a high-production volume (HPV) chemical in the United States. To evaluate its impact on the environment and air quality, its atmospheric reactivity toward the three main tropospheric oxidants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service