Skip to Content
Merck
All Photos(3)

Key Documents

203343

Sigma-Aldrich

Germanium

chips, 99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ge
CAS Number:
Molecular Weight:
72.64
EC Number:
MDL number:
UNSPSC Code:
12141716
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.999% trace metals basis

form

chips

resistivity

53 Ω-cm, 20°C

particle size

≥3 mm

bp

2830 °C (lit.)

mp

937 °C (lit.)

density

5.35 g/mL at 25 °C (lit.)

SMILES string

[Ge]

InChI

1S/Ge

InChI key

GNPVGFCGXDBREM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Streyer et al.
Optics express, 21(7), 9113-9122 (2013-04-11)
We demonstrate strong-to-perfect absorption across a wide range of mid-infrared wavelengths (5-12µm) using a two-layer system consisting of heavily-doped silicon and a thin high-index germanium dielectric layer. We demonstrate spectral control of the absorption resonance by varying the thickness of
Jin Liu et al.
Dalton transactions (Cambridge, England : 2003), 42(14), 5092-5099 (2013-02-13)
In this study, Zn2GeO4 hollow spheres were successfully fabricated by a template-engaged approach using zinc hydroxide carbonate (Zn4CO3(OH)6·H2O, ZHC) spheres as the template. During the hydrothermal process, Zn(2+) dissolved from the surface of the ZHC spheres could rapidly react with
Lu Dai et al.
Nanoscale, 5(3), 971-976 (2012-12-15)
The controllable fabrication of self-scrolling SiGe/Si/Cr helical nanoribbons on Si(111) substrates is investigated. The initial lateral etching profile of the Si(111) substrates shows a 2-fold rotational symmetry using 4% ammonia solution, which provides guidance for initial scrolling of one-end-fixed nanoribbons
Michael Oehme et al.
Optics express, 21(2), 2206-2211 (2013-02-08)
In this paper we investigate the influence of n-type doping in Ge light emitting diodes on Si substrates on the room temperature emission spectrum. The layer structures are grown with a special low temperature molecular beam epitaxy process resulting in
Maurizio Mattesini et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(3), 035601-035601 (2012-12-12)
The magnetic properties, electronic band structure and Fermi surfaces of the hexagonal Cr(2)GeC system have been studied by means of both generalized gradient approximation (GGA) and the +U corrected method (GGA + U). The effective U value has been computed within the

Articles

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service