Skip to Content
Merck
All Photos(2)

Key Documents

GF99720647

Germanium

sheet, 50x50mm, thickness 1.0mm, polycrystalline, 99.999%

Synonym(s):

Germanium, GE003122

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ge
CAS Number:
Molecular Weight:
72.64
MDL number:
UNSPSC Code:
12141716
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.999%

form

sheet

manufacturer/tradename

Goodfellow 997-206-47

resistivity

53 Ω-cm, 20°C

bp

2830 °C (lit.)

mp

937 °C (lit.)

density

5.35 g/mL at 25 °C (lit.)

SMILES string

[Ge]

InChI

1S/Ge

InChI key

GNPVGFCGXDBREM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S H Tao et al.
Regulatory toxicology and pharmacology : RTP, 25(3), 211-219 (1997-06-01)
Germanium-containing dietary supplements became popular in the 1970s in Japan and later in other countries, as elixirs for certain diseases (e.g., cancer and AIDS). Germanium is not an essential element. Its acute toxicity is low. However, at least 31 reported
Yunfei Jiang et al.
Plant, cell & environment, 38(11), 2387-2397 (2015-06-18)
Pea (Pisum sativum L.) is a major legume crop grown in a semi-arid climate in Western Canada, where heat stress affects pollination, seed set and yield. Seed set and pod growth characteristics, along with in vitro percentage pollen germination, pollen
Liqun Zhou et al.
Journal of dentistry, 43(8), 973-980 (2015-06-10)
The present study examined (1) the chemical interaction between three self-etching adhesives and sodium hypochlorite (NaOCl)-deproteinised dentine, and (2) the influence of NaOCl treatment on bond strength of self-etching adhesives with/without adhesive functional monomers to dentine. Caries-free dentine disks (control)
R M Slawson et al.
Plasmid, 27(1), 72-79 (1992-01-01)
Germanium is an inert metal with no known biological function in prokaryotic or eukaryotic organisms. Its toxicity is low compared to that of silver. Germanium is accumulated in certain bacterial strains by either energy-independent passive binding or an energy-dependent mechanism.
Silvie Svarcová et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 132, 514-525 (2014-06-04)
An unambiguous identification of pigments in paint layers of works of art forms a substantial part of the description of a painting technique, which is essential for the evaluation of the work of art including determination of the period and/or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service