573515
Germanium(II) chloride dioxane complex (1:1)
Synonym(s):
Dioxanedichlorogermylene, Germanium dichloride-dioxane
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C4H8Cl2GeO2
CAS Number:
Molecular Weight:
231.65
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid
Recommended Products
form
solid
Quality Level
reaction suitability
reagent type: catalyst
core: germanium
concentration
25.3-35.8% (silver nitrate titration)
SMILES string
Cl[Ge]Cl.C1COCCO1
InChI
1S/C4H8O2.Cl2Ge/c1-2-6-4-3-5-1;1-3-2/h1-4H2;
InChI key
NYMJLNHIEKAQSD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Germanium(II) chloride dioxane complex (1:1) is an organometallic compound used as a reductant in organic synthesis, and to prepare other germanium-based nanoparticles and complexes.
Application
Germanium(II) chloride dioxane complex (1:1) can be used as a precursor:
- To synthesize Germanium oxide nanoparticles.
- To prepare a heterocyclic germylene complex that can be used as an atomic layer deposition precursor to fabricate germanium thin films.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Carc. 2 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Reduction Properties Of Germanium Dichloride With Respect To The Redox-Active Monoiminoacenaphtenone Dpp-Mian}
AN Lukoyanov, et al.
Journal of Structural Chemistry, 64, 288-301 (2023)
From rational design of organometallic precursors to optimized synthesis of core/shell Ge/GeO 2 nanoparticles
D Matioszek, et al.
Dalton Transactions, 44, 7242-7250 (2015)
Synthesis of N-heterocyclic stannylene (Sn (II)) and germylene (Ge (II)) and a Sn (II) amidinate and their application as precursors for atomic layer deposition
Sang Bok Kim, et al.
Chemistry of Materials, 26, 3065-3073 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service