Skip to Content
Merck
All Photos(3)

Key Documents

203351

Sigma-Aldrich

Germanium

powder, −100 mesh, ≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ge
CAS Number:
Molecular Weight:
72.64
EC Number:
MDL number:
UNSPSC Code:
12141716
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

≥99.99% trace metals basis

form

powder

resistivity

53 Ω-cm, 20°C

particle size

−100 mesh

bp

2830 °C (lit.)

mp

937 °C (lit.)

density

5.35 g/mL at 25 °C (lit.)

SMILES string

[Ge]

InChI

1S/Ge

InChI key

GNPVGFCGXDBREM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Flam. Sol. 1 - Repr. 2 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minxian Wu et al.
Physical chemistry chemical physics : PCCP, 15(14), 4955-4964 (2013-02-27)
The electrodeposition of germanium from the ionic liquid 1-butyl-1-methylpyrrolidinium dicyanamide ([BMP][DCA]) and a mixture of [BMP][DCA] and 1-butyl-1-methylpyrrolidinium chloride ([BMP]Cl) was studied using cyclic voltammetry and using an electrochemical quartz crystal microbalance (EQCM). [GeCl4(BuIm)2] (BuIm = N-butylimidazole) was used as
Miroslav Cerný et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(3), 035401-035401 (2012-12-15)
The response of three covalent crystals with a diamond lattice (C, Si and Ge) to uniaxial and a special triaxial (generally nonhydrostatic) loading is calculated from first principles. The lattice deformations are described in terms of variations of bond lengths
Fabian von Rohr et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(7), 075804-075804 (2013-01-25)
The thermoelectric properties between 10 and 300 K and the growth of single crystals of n-type and p-type GeBi(4)Te(7), GeSb(4)Te(7) and Ge(Bi(1-x)Sb(x))(4)Te(7) solid solution are reported. Single crystals were grown by the modified Bridgman method, and p-type behavior was achieved
Philippe Jund et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(3), 035403-035403 (2012-12-15)
We report an ab initio study of the semiconducting Mg(2)X (with X = Si, Ge) compounds and in particular we analyze the formation energies of the different point defects with the aim of understanding the intrinsic doping mechanisms. We find that the
Jian-Qiang Shen et al.
Dalton transactions (Cambridge, England : 2003), 42(16), 5812-5817 (2013-03-05)
An unprecedented organic-inorganic hybrid germanoniobate compound Na4[Cu(en)2(H2O)2]5[Na6Ge8Nb32O108H8(OH)4]·41H2O (1) was synthesized under the hydrothermal condition. In compound 1, the {Nb16} cage containing four {GeO4} tetrahedra in its internal cavity results in a heteropolyniobate anion [H4Ge4Nb16O54(OH)2](10-) ({Ge4Nb16}), which is connected by a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service