Skip to Content
Merck
All Photos(3)

Documents

204544

Sigma-Aldrich

Tellurium

pieces, 99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Te
CAS Number:
Molecular Weight:
127.60
EC Number:
MDL number:
UNSPSC Code:
12141913
eCl@ss:
38040201
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

pieces

resistivity

5.8-33 μΩ-cm, 20°C

bp

990 °C (lit.)

mp

450 °C (lit.)

density

6.24 g/mL at 25 °C (lit.)

SMILES string

[Te]

InChI

1S/Te

InChI key

PORWMNRCUJJQNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tellurium is a p-type narrow band gap semiconducting metalloid with excellentphotoconductive and thermoconductive properties. It is widely as a fundamentalconstituent of photovoltaic solar panels, thermoelectronic devices, andalloys.

Application

Tellurium is an essential component for advanced materials including topological insulators such as thermoelectric materials and telluride nanowires. The form of this material is ideally suited for vacuum deposition techniques such as MBE.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 4 - Lact. - Repr. 1B - Skin Sens. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hridyesh Kumar et al.
Carbohydrate polymers, 97(2), 327-334 (2013-08-06)
The present work describes synthesis of water soluble L-cysteine-functionalized CdTe quantum dots (QDs) with size tunable emission at different time intervals for chitosan based film. The characterization of the synthesized CdTe QDs-chitosan film was made by Fourier transform infrared (FTIR)
E C Stangherlin et al.
Toxicology, 207(2), 231-239 (2004-12-15)
The effect of single maternal subcutaneous (s.c.) injection of 0.12 mg/kg diphenyl ditelluride, (PhTe)2, diluted in canola oil at days 6, 10 or 17 of gestation were evaluated in Wistar rats. The reduction of body weight gain was statistically significant
Edward R T Tiekink
Dalton transactions (Cambridge, England : 2003), 41(21), 6390-6395 (2012-01-19)
Despite being disparaged for their malodorous and toxic demeanour, compounds of selenium, a bio-essential element, and tellurium, offer possibilities as therapeutic agents. Herein, their potential use as drugs, for example, as anti-viral, anti-microbial, anti-inflammatory agents, etc., will be surveyed along
Raymond J Turner et al.
Biotechnology advances, 30(5), 954-963 (2011-09-13)
Here, we overview the most recent advances in understanding the bacterial mechanisms that stay behind the reduction of tellurium oxyanions in both planktonic cells and biofilms. This is a topic of interest for basic and applied research because microorganisms are
Vijay P Singh et al.
Organic letters, 15(24), 6274-6277 (2013-11-28)
In an effort to improve the chain-breaking capacity of the natural antioxidants, an octyltelluro group was introduced next to the phenolic moiety in β- and δ-tocopherol. The new vitamin E analogues quenched peroxyl radicals more efficiently than α-tocopherol and were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service