Skip to Content
Merck
All Photos(2)

Key Documents

224839

Sigma-Aldrich

Bismuth(III) chloride

reagent grade, ≥98%

Synonym(s):

Bismuth trichloride, Trichlorobismuth

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BiCl3
CAS Number:
Molecular Weight:
315.34
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
form:
powder, chunks or granules

grade

reagent grade

Assay

≥98%

form

powder, chunks or granules

reaction suitability

reagent type: catalyst
core: bismuth

bp

447 °C (lit.)

mp

230-232 °C (lit.)

SMILES string

Cl[Bi](Cl)Cl

InChI

1S/Bi.3ClH/h;3*1H/q+3;;;/p-3

InChI key

JHXKRIRFYBPWGE-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Bismuth(III) chloride is an inorganic salt that can be prepared by the reaction of bismuth with chlorine.

Application

Bismuth(III) chloride may be used as a catalyst in the following processes:
  • Alcoholysis, acetolysis and hydrolysis of epoxides to form the corresponding β-alkoxy and β-acetoxy alcohols and diols.
  • Preparation of thiiranes from oxiranes using ammonium thiocyanate.
  • Three component coupling of carbonyl compound, amine and dialkyl phosphite to form α-amino phosphonates.
  • Allylation of secondary benzyl alcohols with allyltrimethylsilane.
  • [4 + 2] cycloaddition between dienes and α-carbonyl ethylenic dienophiles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daintith J
The facts on file dictionary of chemistry, 28-28 (2014)
Bismuth (III) chloride (BiCl3)-An efficient catalyst for mild, regio-and stereoselective cleavage of epoxides with alcohols, acetic Acid and water.
Mohammadpoor-Baltork I.
Synthetic Communications, 30(13), 2365-2374 (2000)
Bismuth (III) chloride-catalyzed direct deoxygenative allylation of substituted benzylic alcohols with allyltrimethylsilane
De SK & Gibbs RA.
Tetrahedron Letters, 46(48), 8345-8350 (2005)
Bismuth (III) Chloride-Catalyzed Three-Component Coupling: Synthesis of a-Amino Phosphonates.
Zhan ZP and Li JP.
Synthetic Communications, 35(19), 2501-2508 (2005)
Bismuth (iii) Chloride Mediated Michaelis-Arbuzov Reaction: A Facile Synthesis of Substituted Arylphosphonates/Phosphinates and Bioactivity Evaluation.
Subramanyam C, et al.
Phosph. Sulfur Relat. Elem. (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service