Merck
  • Markers of mesterolone abuse in sulfate fraction for doping control in human urine.
Markers of mesterolone abuse in sulfate fraction for doping control in human urine.
Journal of mass spectrometry : JMS (2015-12-05)
P Kiousi, Y S Angelis, A G Fragkaki, W Abushareeda, M Alsayrafi, C Georgakopoulos, E Lyris
ABSTRACT
This manuscript describes the direct detection of mesteroloe sulfo-conjugated metabolites by liquid chromatography/quadrupole/time of flight mass spectrometry (LC/Q/TOFMS) with special focus on evaluation of their retrospective detectability and their structure elucidation. A comparison of their long-term detectability, with the mesterolone main metabolite (1α-methyl-5α-androstan-3α-ol-17-one) excreted in glucuronide fraction and detected by gas chromatography/high resolution mass spectrometry (GC/HRMS), is also presented. Studies on mesterolone were performed with samples obtained from two excretion studies after single oral administration of Proviron© by healthy volunteers. Potential sulfate metabolites were detected in post administration samples after liquid-liquid extraction (LLE) with ethyl acetate and LC/TOFMS analysis, in negative mode. Twelve mesterolone sulfate metabolites from the first excretion study and nine from the second were subsequently confirmed by LC/Q/TOFMS. Finally, six mesterolone sulfate metabolites were considered important taking into account their abundance and long-term detectability, encoded as M1, M2, M4, M5, M6 and M7. The proposed mesterolone sulfate metabolites M1, M2, M4 and M5 (excreted as sulfates) have the same retrospectivity with the main mesterolone metabolite, excreted in glucuronide fraction. For metabolite characterization, LC fractionation was performed. The metabolites were identified and characterized by GC/MS, after solvolysis and derivatization. Combined mass spectra data from trimethyl-silyl (TMS), TMS-enolTMS and methoxime-TMS derivatives were taken into account for the characterization of these metabolites. It was concluded that M1 is 1α-methyl-5α-androstan-3β-ol-17 one, M2 is 1α-methyl-5α-androstan-3α-ol-17 one, M4 is 1α-methyl-5a-androstan-3β, 16z-diol-17-one, M5 is 1α-methyl-5α-androstan-17z,4ξ-diol-3one, M6 is 1α-methyl-5α-androstan-3z,6z-diol-17-one and M7 is 4z-hydroxy-1α-methyl-5α-androstan-3,17-dione.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Ammonium iodide, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Supelco
N-Methyl-bis(trifluoroacetamide), for GC derivatization, LiChropur, ≥97.0% (GC)
Sigma-Aldrich
Heptacosafluorotributylamine, liquid
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
1,4-Dithioerythritol, BioReagent, for molecular biology, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, BioXtra, ≥99.0%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0% (RT), BioUltra
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%