Skip to Content
Merck
All Photos(1)

Key Documents

H5262

Sigma-Aldrich

Heptacosafluorotributylamine

liquid

Synonym(s):

PFTBA, Perfluorotributylamine, Tris(nonafluorobutyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
N(CF2CF2CF2CF3)3
CAS Number:
Molecular Weight:
671.09
Beilstein:
1813883
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.25

vapor density

23.3 (vs air)

Quality Level

vapor pressure

1.3 mmHg ( 25 °C)

form

liquid

bp

178 °C (lit.)

density

1.883 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)N(C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C12F27N/c13-1(14,7(25,26)27)4(19,20)10(34,35)40(11(36,37)5(21,22)2(15,16)8(28,29)30)12(38,39)6(23,24)3(17,18)9(31,32)33

InChI key

RVZRBWKZFJCCIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H A Sudhalkar et al.
Retina (Philadelphia, Pa.), 18(5), 460-465 (1998-11-04)
Perfluorocarbon liquids (PFCLs) are useful for floating various materials off the posterior retina during vitreoretinal surgery. It is generally assumed that buoyancy forces restrict this use to objects with specific gravity lower than that of PFCLs. We tested potential intraocular
V Quaresima et al.
Biochimica et biophysica acta, 1366(3), 291-300 (1998-11-14)
Near-infrared spectroscopy was used to determine the relationship between the redox state of mitochondrial cytochrome oxidase CuA and haemoglobin oxygenation in the isoflurane-anaesthetized neonatal pig brain. Adding 7% CO2 to the inspired gases increased the total haemoglobin concentration by 8
V M Knepp et al.
Pharmaceutical research, 15(7), 1090-1095 (1998-08-04)
To identify a suitable nonaqueous, parenterally acceptable suspending vehicle whereby a therapeutic protein is delivered as a stable flowable powder, making it amenable to delivery from sustained delivery systems maintained at body temperature. Formulations of plasma derived Factor IX (pdFIX)
Discordant xenoheart transplantation with perfluorotributylamine/pluronic F-68 stem emulsion.
M Kawauchi et al.
Transplantation proceedings, 30(7), 3831-3832 (1998-12-05)
Nadav Kimelman-Bleich et al.
Biomaterials, 30(27), 4639-4648 (2009-06-23)
A major hurdle to surmount in bone-tissue engineering is ensuring a sufficient oxygen supply to newly forming tissue to avoid cell death or delayed development of osteogenic features. We hypothesized that an oxygen-enriched hydrogel scaffold would enhance tissue-engineered bone formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service