Skip to Content
Merck
All Photos(1)

Key Documents

339741

Sigma-Aldrich

Sulfuric acid

99.999%

Synonym(s):

Dihydrogen sulfate, Oil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22

Agency

suitable for SM 5210

Quality Level

vapor density

<0.3 (25 °C, vs air)

vapor pressure

1 mmHg ( 146 °C)

description

Nominally 95-98% H2SO4

Assay

99.999%

form

viscous liquid

color

clear

pH

1.2 (5 g/L)

bp

~290 °C (lit.)

solubility

water and organic: soluble

density

1.840 g/mL at 25 °C (lit.)

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfuric acid is a strong mineral acid, that is commonly used in organic synthesis as a reagent, catalyst, and solvent. It is also useful to perform several functions, including dehydration, sulfonation, and nitration.

Application

Sulfuric acid is used:
  • As a catalyst in the esterification of acetic and palmitic acid.
  • As a solvent in the synthesis of binary technetium sulfide via the reaction of pertechnetate and hydrogen sulfide.
  • As a dehydrating agent, catalyst, and active reactant in the chemical industry.
  • To prepare silica-sulfuric acid composite, which is used as a catalyst in organic synthesis.

Analysis Note

Purity based on trace metals

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Mechanical Performance of a Biomimetic Nanointerface Made of Multilayered Polyelectrolytes.
Cavelier S, et al.
European Journal of Inorganic Chemistry, 32, 5380-5389 (2012)
Analysis of Malondialdehyde, Chlorophyll Proline, Soluble Sugar, and Glutathione Content in Arabidopsis seedling
Zhang Z, et al.
Bio-protocol, 3(14) (2013)
Esterification of free fatty acids using sulfuric acid as catalyst in the presence of triglycerides
Marchetti JM and Errazu AF
Biomass and Bioenergy, 32(9), 892-895 (2008)
An effective and simple oxygen nanosensor made from MPA-capped water soluble CdTe nanocrystals.
Lau PC,et al.
Nanotechnology, 24(1) (2013)
Silica sulfuric acid and silica chloride as efficient reagents for organic reactions
Salehi P, et al.
Current Organic Chemistry, 10(17), 2171-2189 (2006)

Articles

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Acid and base chart lists the strength of acids and bases (strongest to weakest) in order. Simple to use laboratory reference chart for scientists, researchers and lab technicians.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service