Skip to Content
Merck
All Photos(1)

Key Documents

30743

Sigma-Aldrich

Sulfuric acid

puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

<0.3 (25 °C, vs air)

vapor pressure

1 mmHg ( 146 °C)

description

Nominally 95-98% H2SO4

Assay

95.0-97.0%

form

liquid

impurities

≤1 ppm heavy metals (as Pb)
≤2 ppm KMnO4 red. matter (as O)
≤2 ppm ammonium (NH4)

ign. residue

≤5 ppm

pH

1.2 (5 g/L)

bp

~290 °C (lit.)

density

1.840 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.1 ppm
nitrate (NO3-): ≤0.2 ppm
phosphate (PO43-): ≤0.5 mg/kg

cation traces

Ag: ≤0.02 mg/kg
Al: ≤0.05 mg/kg
As: ≤0.01 ppm
Ba: ≤0.05 mg/kg
Be: ≤0.01 mg/kg
Bi: ≤0.05 mg/kg
Ca: ≤0.2 mg/kg
Cd: ≤0.02 mg/kg
Co: ≤0.01 mg/kg
Cr: ≤0.05 mg/kg
Cu: ≤0.01 mg/kg
Fe: ≤0.1 ppm
Ge: ≤0.05 mg/kg
Hg: ≤5 ppb
K: ≤0.1 mg/kg
Li: ≤0.01 mg/kg
Mg: ≤0.05 mg/kg
Mn: ≤0.01 mg/kg
Mo: ≤0.02 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.02 mg/kg
Sr: ≤0.02 mg/kg
Ti: ≤0.1 mg/kg
Tl: ≤0.05 mg/kg
V: ≤0.01 mg/kg
Zn: ≤0.05 mg/kg
Zr: ≤0.1 mg/kg

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Bauxite for scientific experimentation: The material′s utility extends to experimental applications such as the production of flash setting admixtures, showcasing its versatility in modifying physical properties for various scientific needs (Chen et al., 2024).



Packaging

In PE-bottles;also available PE-bottles black (Material no. ending with PEBL; Glass-bottles (Material no. ending with *-GL), PE canister (Material no. ending with *-H)

Other Notes

The article number 30743-4X2.5L will be discontinued. Please order the single bottle 30743-2.5L which is physically identical with the same exact specifications.
The article number 30743-4X2.5L-M will be discontinued. Please order the single bottle 30743-2.5L-M which is physically identical with the same exact specifications.
The article number 30743-6X1L will be discontinued. Please order the single bottle 30743-1L which is physically identical with the same exact specifications.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of mercury in natural waters and effluents by flameless atomic absorption spectrophotometry.
S H Omang
Analytica chimica acta, 53(2), 415-419 (1971-02-01)
Jing Li et al.
Journal of chromatography. A, 1404, 39-43 (2015-06-09)
The paper reports on a novel method for rapid determination of carbonyl in lignins by headspace gas chromatography (HS-GC). The method involves the quantitative carbonyl reduction for aldehydes in 2min at room temperature or for acetones in 30min at 80°C
Self-Ordering of Cell Arrangement of Anodic Porous Alumina Formed in Sulfuric Acid Solution.
Masuda H, et al.
Journal of the Electrochemical Society, 144(5), L127-L130 (1997)
Investigation of the kinetics and mechanism of the anodic oxidation of aniline in aqueous sulfuric acid solution at a platinum electrode.
Mohilner DM, et al.
Journal of the American Chemical Society, 84(19), 3618-3622 (1962)
Francesco Riccobono et al.
Science (New York, N.Y.), 344(6185), 717-721 (2014-05-17)
Atmospheric new-particle formation affects climate and is one of the least understood atmospheric aerosol processes. The complexity and variability of the atmosphere has hindered elucidation of the fundamental mechanism of new-particle formation from gaseous precursors. We show, in experiments performed

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. 안녕하세요, Sulfuric acid 30743_1L와 30743_1L_GL 간의 차이가 무엇인가요? 감사합니다.

    1 answer
    1. As mentioned in the 'DESCRIPTION' section, the pack size designated as -GL is packaged in glass. Other options listed, without the -GL designation, are packaged in PE bottles.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service