Skip to Content
Merck
All Photos(1)

Key Documents

07208-M

Sigma-Aldrich

Sulfuric acid

Sulfuric acid

puriss., meets analytical specification of Ph. Eur., BP, 95-97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor density

<0.3 (25 °C, vs air)

vapor pressure

1 mmHg ( 146 °C)

description

Nominally 95-98% H2SO4

grade

puriss.

Assay

95-97%

form

solution

quality

meets analytical specification of Ph. Eur., BP

impurities

≤0.0002% KMnO4 red. matter (as O)
≤5 ppm heavy metals (as Pb)

ign. residue

≤0.0005%

bp

~290 °C (lit.)

density

1.840 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1 ppm
nitrate (NO3-): ≤0.0001%

cation traces

As: ≤0.1 ppm
Ca: ≤0.001%
Cd: ≤0.0005%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤2 ppm
K: ≤0.005%
Mg: ≤0.0005%
Mn: ≤0.0005%
Na: ≤0.005%
Pb: ≤0.0005%
Zn: ≤0.0005%

suitability

complies for appearance of solution

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sulfuric acid is commonly used in regulated and highly demand lab applications such as quality control and production. In organic synthesis, sulfuric acid can be used as a dehydrating agent, catalyst, and active reactant.

Packaging

In PE-bottles; also available in Glass-bottles (Material no. ending with *-GL), PE canister (Material no. ending with *-H)

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Li et al.
Journal of chromatography. A, 1404, 39-43 (2015-06-09)
The paper reports on a novel method for rapid determination of carbonyl in lignins by headspace gas chromatography (HS-GC). The method involves the quantitative carbonyl reduction for aldehydes in 2min at room temperature or for acetones in 30min at 80°C
G Carmona et al.
Scandinavian journal of medicine & science in sports, 25(6), e547-e557 (2014-12-03)
Muscle damage induced by inertial exercise performed on a flywheel device was assessed through the serum evolution of muscle enzymes, interleukin 6, and fiber type-specific sarcomere proteins such as fast myosin (FM) and slow myosin (SM). We hypothesized that a
Babak Anasori et al.
ACS nano, 9(10), 9507-9516 (2015-07-25)
The higher the chemical diversity and structural complexity of two-dimensional (2D) materials, the higher the likelihood they possess unique and useful properties. Herein, density functional theory (DFT) is used to predict the existence of two new families of 2D ordered
Andreas Recke et al.
Journal of immunology (Baltimore, Md. : 1950), 193(4), 1600-1608 (2014-07-16)
Subepidermal autoimmune blistering dermatoses (AIBD) are prototypic autoantibody-mediated diseases. In epidermolysis bullosa acquisita (EBA), an autoimmune disease with severe and chronic skin blistering, autoantibodies are directed against type VII collagen. IgG is the predominant autoantibody isotype of EBA, the pathogenicity
Ban Xiong Tan et al.
Oncotarget, 5(4), 933-943 (2014-03-25)
Although p53 is found mutated in almost 50% of all cancers, p53 mutations in leukaemia are relatively rare. Acute myeloid leukaemia (AML) cells employ other strategies to inactivate their wild type p53 (WTp53), like the overexpression of the p53 negative

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service