Skip to Content
Merck
All Photos(4)

Documents

71690

Sigma-Aldrich

Sodium hydroxide

puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets

Synonym(s):

‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

vapor density

>1 (vs air)

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

Assay

≥98.0% (T)

form

pellets

technique(s)

cell culture | plant: suitable

impurities

≤0.0003% total nitrogen (N)
≤0.0005% silicon dioxide (SiO2)
≤1.0% sodium carbonate (Na2CO3)

mp

318 °C (lit.)

solubility

water: soluble 1,260 g/L at 20 °C

density

2.13 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤5 mg/kg

cation traces

Ag: ≤5 mg/kg
Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤5 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Hg: ≤0.1 mg/kg
K: ≤200 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Tl: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium hydroxide (NaOH) is an inorganic base. It participates in the oxidation of glycerol catalyzed by Au/charcoal or Au/graphite. It also participates in solvent-free aldol condensation reaction.

Application

Sodium hydroxide may be employed for the direct oxidation of sodium borohydride, which was investigated by cyclic and linear voltammetry, chronoamperometry and chronopotentiometry. It may be used to compose citrate buffer and borate buffers.
It may be used in the synthesis of following compounds:
  • alkylaryl amidosulfobetaines
  • (3-[ 4-teroctylphenoxyethoxyethoxyethyl] dimethylammonio propane sulfonate (XOSB)
  • trisulfobetaines (TriSBn)

Other Notes

Reagent for the decomposition of silicates by fusion

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Bock
Aufschlussmethoden der anorg. & org. Chemie (1972)
Z. Sulcek et al.
Methods of Decomposition in Inorganic Analysis (1989)
Chemoselective, solvent-free aldol condensation reaction.
Raston CL and Scott JL.
Green Chemistry, 2(2), 49-52 (2000)
M A Roberts et al.
Analytical chemistry, 69(11), 2035-2042 (1997-06-01)
This report describes a UV laser photoablation method for the production of miniaturized liquid-handling systems on polymer substrate chips. The fabrication of fluid channel and reservoir networks is accomplished by firing 200 mJ pulses from an UV excimer laser at
Kinetics of sodium borohydride direct oxidation and oxygen reduction in sodium hydroxide electrolyte: Part I. BH4- electro-oxidation on Au and Ag catalysts.
Chatenet M, et al.
Electrochimica Acta, 51(25), 5459-5467 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service