Skip to Content
Merck
All Photos(4)

Key Documents

901915

Sigma-Aldrich

Sodium hydroxide

anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets

Synonym(s):

‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
Assay:
≥98%
grade:
anhydrous
reagent grade
form:
pellets
solubility:
water: soluble 1260 g/L at 20 °C
Pricing and availability is not currently available.

grade

anhydrous
reagent grade

vapor density

>1 (vs air)

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

product line

Redi-Dri

Assay

≥98%

form

pellets

quality

free-flowing

impurities

≤1.0% sodium carbonate

pH

14 (20 °C, 50 g/L)

mp

318 °C (lit.)

solubility

water: soluble 1260 g/L at 20 °C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Redi-Dri technology eliminates clumping and the need for complicated procedures to keep your salts free-flowing. Keeping your salts dry helps improve your results by giving accurate measurements and easier-to-handle salts and buffers. Rather than drying your salts in an oven or desiccator or breaking them apart manually, Redi-Dri ensures that your salts will remain easy-to-use for months without worrying about anti-caking agents. Even after repeated openings you can be assured that your results are always accurate. Redi-Dri saves you time, energy and money by keeping your salts usable for longer!

Application

  • Sodium hydroxide has been used in the staining procedure of cells.[1]
  • It has been used in the procedure of decellularizing human amniotic membrane for cell culture.[2]
  • It has been used for chemical inactivation of fluorophores in immunostained cells.[3]

Features and Benefits

  • Increased efficiency – Eliminate time and effort of loosening hardened powders
  • Safety – Avoid risks of using tools of force to break up clumps
  • Economy – Faster preparation and solvation increase productivity and reduce costs
  • Assured quality – Excellent, expert-tested quality with no anti-caking agents
  • Flexible volumes – Available from research amounts to scale-up quantities

Packaging

Bulk packages

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple alkaline method for decellularizing human amniotic membrane for cell culture.
Saghizadeh M et al.
PLoS ONE, 8, e79632-e79632 (2013)
BDNF Pretreatment of Human Embryonic-Derived Neural Stem Cells Improves Cell Survival and Functional Recovery After Transplantation in Hypoxic-Ischemic Stroke.
Rosenblum S et al.
Cell Transplantation, 24, 2449-2449 (2015)
Jia-Ren Lin et al.
Nature communications, 6, 8390-8390 (2015-09-25)
Single-cell analysis reveals aspects of cellular physiology not evident from population-based studies, particularly in the case of highly multiplexed methods such as mass cytometry (CyTOF) able to correlate the levels of multiple signalling, differentiation and cell fate markers. Immunofluorescence (IF)
Jesper Hjortnaes et al.
Advanced healthcare materials, 4(1), 121-130 (2014-06-25)
Three dimensional (3D) hydrogel platforms are powerful tools, providing controllable, physiologically relevant microenvironments that could aid in understanding how various environmental factors direct valvular interstitial cell (VIC) phenotype. Continuous activation of VICs and their transformation from quiescent fibroblast to activated
Richard J Travers et al.
Blood, 124(22), 3183-3190 (2014-09-10)
Polyphosphate (polyP) is secreted by activated platelets and has been shown to contribute to thrombosis, suggesting that it could be a novel antithrombotic target. Previously reported polyP inhibitors based on polycationic substances, such as polyethylenimine, polyamidoamine dendrimers, and polymyxin B

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

1–3 of 3 Questions  
  1. How does Redi-Dri™ help keep salts and buffers free-flowing?

    1 answer
    1. Redi-Dri™ products are specially handled and the bottle is specially designed to ensure that the product will remain dry without adding impurities or additives to the product. Ensuring that you receive the highest quality material that is easy to handle.

      Helpful?

  2. How long will material remain free-flowing after it is opened?

    1 answer
    1. While the exact amount of time your material will remain free-flowing can vary, this largely depends on the ambient storage conditions and how many times it is opened.

      Helpful?

  3. Are there any special storage conditions that must be maintained?

    1 answer
    1. No, Redi-Dri™ products can be stored under the same conditions as the equivalent non-Redi-Dri™ products.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service