Skip to Content
Merck
All Photos(1)

Key Documents

I2764

Sigma-Aldrich

ML-7

powder

Synonym(s):

1-(5-Iodonaphthalene-1-sulfonyl)-1H-hexahydro-1,4-diazepine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17IN2O2S · HCl
CAS Number:
Molecular Weight:
452.74
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC)

form

powder

color

white

solubility

ethanol: water (1:1): 10 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

Cl.Ic1cccc2c(cccc12)S(=O)(=O)N3CCCNCC3

InChI

1S/C15H17IN2O2S.ClH/c16-14-6-1-5-13-12(14)4-2-7-15(13)21(19,20)18-10-3-8-17-9-11-18;/h1-2,4-7,17H,3,8-11H2;1H

InChI key

KDDALCDYHZIZMH-UHFFFAOYSA-N

Gene Information

Application

ML-7 has been used as a myosin light chain kinase (MLCK) inhibitor in various experiments.

Biochem/physiol Actions

Selective myosin light chain kinase inhibitor.

Features and Benefits

This compound is featured on the Ca/CaMKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ziad Al Nabhani et al.
Inflammatory bowel diseases, 23(7), 1109-1119 (2017-05-19)
Crohn's disease (CD) pathogenesis is multifactorial involving genetic and environmental factors. Loss of function mutations in the nucleotide oligomerization domain 2 (NOD2) gene are the main genetic risk factor for CD. Like patients with CD, Nod2 mice are characterized by
Zasp/Cypher internal ZM-motif containing fragments are sufficient to co-localize with alpha-actinin?Analysis of patient mutations
Klaavuniemi T and Ylanne J
Experimental Cell Research, 312(8), 1299-1311 (2006)
Kristopher E Kubow et al.
Scientific reports, 7(1), 14380-14380 (2017-11-01)
Contact guidance-cell polarization by anisotropic substrate features-is integral to numerous physiological processes; however the complexities of its regulation are only beginning to be discovered. In particular, cells polarize to anisotropic features under non-muscle myosin II (MII) inhibition, despite MII ordinarily
M Makishima et al.
FEBS letters, 287(1-2), 175-177 (1991-08-05)
Inhibitors of myosin light chain kinase, 1-(5-chloronaphthalene-1-sulfonyl)-1H-hexahydro-1,4-diazepine hydrochloride (ML-9) and 1-(5-iodonaphthalene-1-sulfonyl)-1H-hexahydro-1,4-diazepine hydrochloride (ML-7), induced Nitroblue tetrazolium reducing activity, lysozyme activity and morphological maturation of human monoblastic U937, THP-1 and promyelocytic HL-60 cells, but not of erythroblastic K562 cells. However, three
Inhibition of myosin light chain kinase can be targeted for the development of new therapies against herpes simplex virus type-1 infection
Antoine TE and Shukla D
Antiviral Therapy, 19, 15-29 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service