Skip to Content
Merck
All Photos(4)

Key Documents

L7260

Sigma-Aldrich

Oleoyl-L-α-lysophosphatidic acid sodium salt

≥98%, solid

Synonym(s):

1-Oleoyl-sn-glycerol 3-phosphate sodium salt, 3-sn-Lysophosphatidic acid, 1-oleoyl sodium salt, LPA sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H41O7P · xNa+
CAS Number:
Molecular Weight:
436.52 (free acid basis)
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98%

form

solid

color

white

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(O)=O)([H])COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C21H41O7P.Na.H/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(23)27-18-20(22)19-28-29(24,25)26;;/h9-10,20,22H,2-8,11-19H2,1H3,(H2,24,25,26);;/b10-9+;;

InChI key

CHJOEWDRTPWOCY-TTWKNDKESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lysophosphatidic acid (LPA) is an essential metabolite for membrane biosynthesis. LPA interacts with the G protein-coupled receptors (GPCRs), called the LPA receptor and mediates signaling. Oleoyl-L-α-lysophosphatidic acid activates the receptor LPA4. In keratinocytes, oleoyl-L-α-lysophosphatidic acid promotes growth to some extent.

Application

Oleoyl-L-α-lysophosphatidic acid sodium salt has been used:
  • for the activation of Ras homolog gene family, member A (RhoA) and expression of claudin-1 in human breast cancer epithelial cell line
  • in RH7777 cells for cyclic adenosine monophosphate (cAMP) accumulation assay and calcium mobilisation assay
  • in vitro luciferase assay and live-cell imaging

Biochem/physiol Actions

Endogenous agonist for LPA1 and LPA2 receptors. LPA does not induce angiogenesis, but has effects on endothelial cell physiology that are similar to those of sphingosine 1-phosphate. Induces cell migration of cancer and non-cancer cells.

Features and Benefits

This compound is featured on the Lysophospholipid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Hygroscopic, photosensitive

Preparation Note

Prepared by the action of phospholipase A on L-α-phosphatidic acid, dioleoyl.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tracee Scalise Panetti
Biochimica et biophysica acta, 1582(1-3), 190-196 (2002-06-19)
This review discusses multiple effects of sphingosine 1-phosphate (S1P) and lysophosphatidic acid (LPA) on endothelial cells and proposes that S1P and LPA are important regulators of the vascular system. Two physiologic sources of S1P and LPA are platelets and lipoproteins.
A Gohla et al.
The Journal of biological chemistry, 273(8), 4653-4659 (1998-03-21)
Lysophosphatidic acid (LPA) utilizes a G-protein-coupled receptor to activate the small GTP-binding protein Rho and to induce rapid remodeling of the actin cytoskeleton. We studied the signal transduction from LPA receptors to Rho activation. Analysis of the G-protein-coupling pattern of
Identification of compounds acting as negative allosteric modulators of the LPA 1 receptor
Ellery J, et al.
European Journal of Pharmacology (2018)
Phosphatidic acid has a potential to promote hair growth in vitro and in vivo, and activates mitogen-activated protein kinase/extracellular signal-regulated kinase kinase in hair epithelial cells
Takahashi T, et al.
The Journal of Investigative Dermatology, 121(3), 448-456 (2003)
F N van Leeuwen et al.
Biochemical Society transactions, 31(Pt 6), 1209-1212 (2003-12-04)
LPA (lysophosphatidic acid), the simplest of al glycerophospholipids, is a potent inducer of cell proliferation, migration and survival. It does so by activating its cognate G-protein-coupled receptors, four of which have been identified. LPA receptors couple to at least three

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service