Skip to Content
Merck
All Photos(1)

Key Documents

475880

Sigma-Aldrich

ML-7, Hydrochloride

A cell-permeable, potent, reversible, ATP-competitive, and selective inhibitor of myosin light chain kinase (Ki = 300 nM).

Synonym(s):

ML-7, Hydrochloride, 1-(5-Iodonaphthalene-1-sulfonyl)homopiperazine, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17IN2O2S · xHCl
CAS Number:
Molecular Weight:
416.28 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

>98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

50% ethanol: 4.5 mg/mL
DMSO: 4.5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Ic1c2c(c(ccc2)[S](=O)(=O)N3CCNCCC3)ccc1

InChI

1S/C15H17IN2O2S/c16-14-6-1-5-13-12(14)4-2-7-15(13)21(19,20)18-10-3-8-17-9-11-18/h1-2,4-7,17H,3,8-11H2

InChI key

GEHJIACZUFWBTK-UHFFFAOYSA-N

General description

A cell-permeable, potent, reversible, ATP-competitive, and selective inhibitor of myosin light chain kinase (Ki = 300 nM). Inhibits protein kinase A (Ki = 21 µM) and protein kinase C (Ki = 42 µM) at much higher concentrations.
A cell-permeable, potent, reversible, ATP-competitive, and selective inhibitor of myosin light chain kinase (Ki = 300 nM). Inhibits protein kinase A (Ki = 21 µM)and protein kinase C (Ki = 42 µM) at much higher concentrations.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Myosin light chain kinase (Mlck)
Product competes with ATP.
Reversible: yes
Target Ki: 300 nM against myosin light chain kinase

Warning

Toxicity: Standard Handling (A)

Preparation Note

Further dilute with aqueous buffer just prior to use.

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 6 months at 4°C.

Other Notes

Krarup, T., et al. 1998. Am. J. Physiol. 275, C239.
Saitoh, M., et al. 1987. J. Biol. Chem.262, 7796.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haikel Dridi et al.
The Journal of clinical investigation, 132(4) (2022-02-16)
Patients with heart failure (HF) have augmented vascular tone, which increases cardiac workload, impairing ventricular output and promoting further myocardial dysfunction. The molecular mechanisms underlying the maladaptive vascular responses observed in HF are not fully understood. Vascular smooth muscle cells
Hélène Doerflinger et al.
Current biology : CB, 32(2), 374-385 (2021-12-03)
The Drosophila anterior-posterior axis is specified at mid-oogenesis when the Par-1 kinase is recruited to the posterior cortex of the oocyte, where it polarizes the microtubule cytoskeleton to define where the axis determinants, bicoid and oskar mRNAs, localize. This polarity
Hendrik Schürmann et al.
Journal of cell science, 135(10) (2022-05-28)
Podosomes are mechanosensitive protrusive actin structures that are prominent in myeloid cells, and they have been linked to vascular extravasation. Recent studies have suggested that podosomes are hierarchically organized and have coordinated dynamics on the cell scale, which implies that
Mitchell J George et al.
International journal of molecular sciences, 22(12) (2021-07-03)
Myosin Light Chain (MLC) regulates platelet contraction through its phosphorylation by Myosin Light Chain Kinase (MLCK) or dephosphorylation by Myosin Light Chain Phosphatase (MLCP). The correlation between platelet contraction force and levels of MLC phosphorylation is unknown. We investigate the
Yi-Ting Yeh et al.
PLoS pathogens, 18(3), e1010309-e1010309 (2022-03-23)
The eggs of the parasitic blood fluke, Schistosoma, are the main drivers of the chronic pathologies associated with schistosomiasis, a disease of poverty afflicting approximately 220 million people worldwide. Eggs laid by Schistosoma mansoni in the bloodstream of the host

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service