Skip to Content
Merck
All Photos(3)

Key Documents

B0560

Sigma-Aldrich

(−)-Blebbistatin

≥98% (HPLC), solid, cell cycle inhibitor

Synonym(s):

1-Phenyl-1,2,3,4-tetrahydro-4-hydroxypyrrolo[2.3-b]-7-methylquinolin-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N2O2
CAS Number:
Molecular Weight:
292.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

(−)-Blebbistatin, solid, synthetic

form

solid

solubility

DMSO: 5 mg/mL
chloroform: soluble(lit.)
ethyl acetate: soluble(lit.)
methanol: soluble(lit.)
methylene chloride: soluble(lit.)

storage temp.

−20°C

SMILES string

Cc1ccc2N=C3N(CC[C@@]3(O)C(=O)c2c1)c4ccccc4

InChI

1S/C18H16N2O2/c1-12-7-8-15-14(11-12)16(21)18(22)9-10-20(17(18)19-15)13-5-3-2-4-6-13/h2-8,11,22H,9-10H2,1H3/t18-/m1/s1

InChI key

LZAXPYOBKSJSEX-GOSISDBHSA-N

General description

(-)-Blebbistatin, a bioactive small molecule, is a selective inhibitor of skeletal muscle and non-muscle myosin II isoforms.The mechanism of action of blebbistatin, a myosin II ATPase inhibitor, does not influence other members of the myosin superfamily, such as I, V, and X.Blebbistatin is highly light sensitive, and photobleaching can reverse its effect completely. Blebbistatin is a commonly used as excitation-contraction uncoupling agent in cardiovascular physiology. Blebbistatin controls the inhibitory effect of thrombin on intercellular Ca2+ wave propagation. Blebbistatin reduces cardiac contractility by stabilizing the OFF state of the thick filament.Blebbistatin’s selectivity and affinity for multiple class II myoisin make it an attractive candidate for use in cell motility research and muscle physiology.

Application

(−)-Blebbistatin has been used to investigate the effect on cardiac contractility and effect of thrombin on actomyosin contractility

Biochem/physiol Actions

(-)-Blebbistatin is a cell cycle inhibitor; selective inhibitor of non-muscle myosin II.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The myosin II ATPase inhibitor blebbistatin prevents thrombin-induced inhibition of intercellular calcium wave propagation in corneal endothelial cells
Ponsaerts R, et al.
Investigative Ophthalmology & Visual Science, 49(11), 4816-4827 (2008)
Blebbistatin: use as inhibitor of muscle contraction
Farman GP, et al.
Pflugers Archiv : European Journal of Physiology, 455(6), 995-1005 (2008)
Elena Kassianidou et al.
Cell reports, 27(6), 1897-1909 (2019-05-09)
Three-dimensional matrices often contain highly structured adhesive tracks that require cells to turn corners and bridge non-adhesive areas. Here, we investigate these complex processes using micropatterned cell adhesive frames. Spreading kinetics on these matrices depend strongly on initial adhesive position
Omecamtiv mercabil and blebbistatin modulate cardiac contractility by perturbing the regulatory state of the myosin filament
Kampourakis T, et al.
The Journal of Physiology, 596(1), 31-46 (2018)
Valerie C Coffman et al.
Molecular biology of the cell, 20(24), 5195-5210 (2009-10-30)
Two prevailing models have emerged to explain the mechanism of contractile-ring assembly during cytokinesis in the fission yeast Schizosaccharomyces pombe: the spot/leading cable model and the search, capture, pull, and release (SCPR) model. We tested some of the basic assumptions

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service