Skip to Content
Merck
All Photos(4)

Key Documents

M1404

Sigma-Aldrich

Nocodazole

≥99% (TLC), powder

Synonym(s):

Methyl N-(5-thenoyl-2-benzimidazolyl)carbamate, Methyl [5-(2-thienylcarbonyl)-1H-benzimidazol-2-yl]carbamate, Oncodazole, R 17934, [5-(2-Thienylcarbonyl)-1H-benzimidazol-2-yl]-carbamic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11N3O3S
CAS Number:
Molecular Weight:
301.32
Beilstein:
1085978
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥99% (TLC)

form

powder

mp

300 °C (dec.)

solubility

DMSO: soluble 10 mg/mL (may require heating)
H2O: insoluble

storage temp.

2-8°C

SMILES string

COC(=O)Nc1nc2cc(ccc2[nH]1)C(=O)c3cccs3

InChI

1S/C14H11N3O3S/c1-20-14(19)17-13-15-9-5-4-8(7-10(9)16-13)12(18)11-3-2-6-21-11/h2-7H,1H3,(H2,15,16,17,19)

InChI key

KYRVNWMVYQXFEU-UHFFFAOYSA-N

Gene Information

human ... TUBB(203068)

Looking for similar products? Visit Product Comparison Guide

General description

Nocodazole (NZO) is an experimental benzimidazole-based agent that targets both protein kinases and microtubules. It serves as a lead compound in the quest for new colchicine binding site inhibitors (CBSIs). This co-crystallized ligand acts as a high-affinity ligand for cancer-related kinases ABL, c-KIT, BRAF, and MEK, and functions as a rapidly-reversible inhibitor of microtubule polymerization.

Application

Nocodazole has been used to induce microtubule depolymerization in mouse melanoma B16-F1 cells. It has also been used to treat A549 cells for mitotic arrest.

Biochem/physiol Actions

Nocodazole is an antimitotic agent that disrupts microtubules by binding to β−tubulin and preventing formation of one of the two interchain disulfide linkages, thus inhibiting microtubule dynamics, disruption of mitotic spindle function, and fragmentation of the Golgi complex. Nocodazole arrests the cell cycle at G2/M phase and also prevents phosphorylation of the T cell antigen receptor and inhibits its activity. Nocodazole stimulates the intrinsic GTPase activity of tubulin and activates the JNK/SAPK signaling pathway and induces apoptosis in several normal and tumor cell lines. Nocodazole has been shown to enhance CRISPR homology-directed repair (HDR) efficiency and increase Cas9-mediated editing frequencies.

Physical form

Color white to faint yellow and pink

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muralidharan Mani et al.
Biochimica et biophysica acta. Molecular cell research, 1866(9), 1463-1474 (2019-06-15)
The perinuclear stacks of the Golgi apparatus maintained by dynamic microtubules are essential for cell migration. Activation of Akt (protein kinase B, PKB) negatively regulates glycogen synthase kinase 3β (GSK3β)-mediated tau phosphorylation, which enhances tau binding to microtubules and microtubule
Yuan He et al.
Journal of virology, 84(24), 12832-12840 (2010-09-24)
Many viruses interact with the host cell division cycle to favor their own growth. In this study, we examined the ability of influenza A virus to manipulate cell cycle progression. Our results show that influenza A virus A/WSN/33 (H1N1) replication
Microtubule dynamics alter the interphase nucleus
Gerlitz G, et al.
Cellular and Molecular Life Sciences, 70, 1255-1268 (2013)
Revisiting activity of some nocodazole analogues as a potential anticancer drugs using molecular docking and DFT calculations
Khattab M and Al-Karmalawy AA
Frontiers in Chemistry, 9, 628398-628398 (2021)
Alessandra Pisciottani et al.
Cells, 8(7) (2019-07-10)
Abscission is the final step of cell division, mediating the physical separation of the two daughter cells. A key player in this process is the microtubule-severing enzyme spastin that localizes at the midbody where its activity is crucial to cut

Articles

Modulation of homology-directed repair (HDR) within the context of CRISPR-genome editing has led to the identification of small molecules that enhance CRISPR-mediated HDR efficiency in various cell types.

Modulation of homology-directed repair (HDR) within the context of CRISPR-genome editing has led to the identification of small molecules that enhance CRISPR-mediated HDR efficiency in various cell types.

Modulation of homology-directed repair (HDR) within the context of CRISPR-genome editing has led to the identification of small molecules that enhance CRISPR-mediated HDR efficiency in various cell types.

Modulation of homology-directed repair (HDR) within the context of CRISPR-genome editing has led to the identification of small molecules that enhance CRISPR-mediated HDR efficiency in various cell types.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service