Skip to Content
Merck
All Photos(1)

Key Documents

123072

Sigma-Aldrich

Luminol

97%

Synonym(s):

3-Aminophthalhydrazide, 5-Amino-2,3-dihydro-1,4-phthalazinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N3O2
CAS Number:
Molecular Weight:
177.16
Beilstein:
383929
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

97%

mp

>300 °C (lit.)

SMILES string

Nc1cccc2C(=O)NNC(=O)c12

InChI

1S/C8H7N3O2/c9-5-3-1-2-4-6(5)8(13)11-10-7(4)12/h1-3H,9H2,(H,10,12)(H,11,13)

InChI key

HWYHZTIRURJOHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Luminol is a very efficient and commonly used chemiluminiscent compound in condensed phase. When basic solutions of luminol containing oxygen are treated with an oxidizing agent, light is produced. Oxidising agents capable of initiating this particular reaction are Nitrogen dioxide, PAN, hydrogen peroxide, ozone and other atmospheric oxidants.[1]

Application

  • As a chemiluminiscent substance. Used to measure opsonic and phagocytic function.[2]
  • As a biological sensor. Used to detect the polymorphonuclear leukocytes (PMNL) response in a patient with a myeloperoxidase (MPO) deficiency.[3]
  • Used as a forensic test for blood.[4]
Luminol has been used in a protocol to study the immune-related responses in Arabidopsis thaliana via luminol/peroxidase-based assay.[5]

Features and Benefits

  • Very effective chemiluminiscent compound.
  • Can detect sub-part per billion levels of metal ions with simple and inexpensive instrumentation.[6]
Used for chemiluminescence analysis of, for example, metal cations, blood, and glucococorticoids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C S Easmon et al.
Immunology, 41(1), 67-74 (1980-09-01)
Luminol-dependent chemiluminescence produced by peripheral blood phagocytic cells was measured using a light detecting instrument, the Luminometer 1250 (LKB Wallac). Although less sensitive than liquid scintillation counters, the Luminometer can measure chemiluminescence effectively and detect experimentally induced and clinical variations
T I Quickenden et al.
Luminescence : the journal of biological and chemical luminescence, 16(4), 295-298 (2001-08-21)
A wide range of domestic and industrial substances that might be mistaken for haemoglobin in the forensic luminol test for blood were examined. The substances studied were in the categories of vegetable or fruit pulps and juices; domestic and commercial
Mechanism of cobalt catalysis of luminol chemiluminescence
Burdo, Timothy G., and W. Rudolf Seitz
Analytical Chemistry, 47, 1639-1643 (1975)
C Dahlgren et al.
Infection and immunity, 39(2), 736-741 (1983-02-01)
When polymorphonuclear leukocytes (PMNL) and soluble or particulate matter interact, the cells produce chemiluminescence, linked to activation of the oxidative metabolism of the cells. PMNL isolated from a patient with a myeloperoxidase deficiency were found to produce almost no luminol-dependent
Fast gas chromatography with luminol chemiluminescence detection for the simultaneous determination of nitrogen dioxide and peroxyacetyl nitrate in the atmosphere
Marley, Nancy A., et al.
The Review of Scientific Instruments, 75, 4595-4605 (2004)

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service