198285
Nigrosin
certified by the Biological Stain Commission
Synonym(s):
Acid black 2, Nigrosin water soluble
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
CAS Number:
Colour Index Number:
50420
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47
Recommended Products
grade
certified by the Biological Stain Commission
Quality Level
form
solid
solubility
H2O: 0.1%
H2O: soluble
λmax
570 nm
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
Suitability
Certified for use in Dorner′s spore stain. Used as a negative stain in place of India ink for spirochetes, bacteria, protozoa and fungi.
Used as a negative stain in place of India ink for spirochetes, bacteria, protozoa and fungi.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chris Campbell et al.
Biomedical optics express, 11(10), 5800-5813 (2020-11-06)
Hemozoin (Hz) is a crystal by-product of hemoglobin consumption by malaria parasites. There are currently no in vivo deep tissue sensing methods that can quantify Hz presence noninvasively, which would be advantageous for malaria research and treatment. In this work
Lina Hacker et al.
Journal of biomedical optics, 25(9) (2020-09-06)
Photoacoustic imaging (PAI) enables the detection of blood hemoglobin (HB) concentration and oxygenation (sO2) with high contrast and resolution. Despite the heavy use of photoacoustically determined total hemoglobin (THb) and oxygenation (sO2) biomarkers in PAI research, their relationship with underlying
Marcel Gehrung et al.
Journal of biomedical optics, 24(12), 1-11 (2019-10-19)
Photoacoustic tomography (PAT) is intrinsically sensitive to blood oxygen saturation (sO2) in vivo. However, making accurate sO2 measurements without knowledge of tissue- and instrumentation-related correction factors is extremely challenging. We have developed a low-cost flow phantom to facilitate validation of
Emmanuel I Odongo-Aginya et al.
African health sciences, 7(1), 33-36 (2007-07-03)
The Kato-Katz thick smear technique is the standard technique recommended by the World Health Organisation for the quantitative diagnosis of Schistosoma mansoni and other intestinal helminth infections. The major problem of the technique is that a few hours after the
S C Esteves et al.
Fertility and sterility, 66(5), 798-804 (1996-11-01)
To determine whether the hypo-osmotic swelling test can assess viability of frozen spermatozoa. Fresh semen specimens were divided into two aliquots: one was not processed and the other was processed by swim-up. Both aliquots were then cryopreserved by the liquid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service