Skip to Content
Merck
All Photos(2)

Documents

767921

Sigma-Aldrich

1,3,5-Triscarboxyphenylethynylbenzene

greener alternative

≥95%

Synonym(s):

4,4′,4′′-(1,3,5-benzenetriyltri-2,1-ethynediyl)trisbenzoic acid, 4,4′,4′′-[benzene-1,3,5-triyl-tris(ethyne-2,1-diyl)]tribenzoic acid, BTE, H3BTE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H18O6
CAS Number:
Molecular Weight:
510.49
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

≥95%

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

222-223 (decomposition)

greener alternative category

InChI

1S/C33H18O6/c34-31(35)28-13-7-22(8-14-28)1-4-25-19-26(5-2-23-9-15-29(16-10-23)32(36)37)21-27(20-25)6-3-24-11-17-30(18-12-24)33(38)39/h7-21H,(H,34,35)(H,36,37)(H,38,39)

InChI key

NIJMZBWVSRWZFZ-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Expanded benzene tricarboxylate MOF linker used to synthesize MOFs with ultra high porosity; example MOF-180 and MOF-210. These MOFs show promising applications in gas adsorption; seperations; storage and more.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroyasu Furukawa et al.
Science (New York, N.Y.), 329(5990), 424-428 (2010-07-03)
Crystalline solids with extended non-interpenetrating three-dimensional crystal structures were synthesized that support well-defined pores with internal diameters of up to 48 angstroms. The Zn4O(CO2)6 unit was joined with either one or two kinds of organic link, 4,4',4''-[benzene-1,3,5-triyl-tris(ethyne-2,1-diyl)]tribenzoate (BTE), 4,4',44''-[benzene-1,3,5-triyl-tris(benzene-4,1-diyl)]tribenzoate (BBC)
Kobayashi; N.; et al.
Journal of Materials Chemistry, 18, 1037-1037 (2008)
Nianyong Zhu et al.
Chemical communications (Cambridge, England), 48(30), 3638-3640 (2012-03-07)
Tri- and pentanuclear, kinetically stable SBUs were exploited for the preparation of the novel MOFs [Zn(3)(BTEB)(2)(DMF)(2)] and (Me(2)NH(2))[Zn(5)(BTEB)(3)(μ(3)-OH)(2)(DMF)(2)]. The applied synthetic approach results in topologies that are stabilised by tritopic benzene-trisethynylbenzoic acid (BTEB) linkers giving rise to chiral frameworks with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service