Skip to Content
Merck
All Photos(3)

Documents

686859

Sigma-Aldrich

1,3,5-Tris(4-carboxyphenyl)benzene

greener alternative

≥98%, ≤20 wt. % solvent

Synonym(s):

4,4′,4′′,-Benzene-1,3,5-triyl-tris(benzoic acid), H3BTB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H18O6
CAS Number:
Molecular Weight:
438.43
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

≥98%

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤20 wt. % solvent

mp

322-327 °C

greener alternative category

SMILES string

OC(=O)c1ccc(cc1)-c2cc(cc(c2)-c3ccc(cc3)C(O)=O)-c4ccc(cc4)C(O)=O

InChI

1S/C27H18O6/c28-25(29)19-7-1-16(2-8-19)22-13-23(17-3-9-20(10-4-17)26(30)31)15-24(14-22)18-5-11-21(12-6-18)27(32)33/h1-15H,(H,28,29)(H,30,31)(H,32,33)

InChI key

SATWKVZGMWCXOJ-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

1,3,5-Tris(4-carboxyphenyl)benzene (H3BTB) is a star-shaped two-dimensional molecule that forms a self-assembled monolayer (SAM) on a variety of substrates. It can be used as a tritopic bridging ligand that facilitates the functionalization of polyoxometalate-based metal organic frameworks (MOFs) for potential usage in gas storage, gas separation, and catalysis.
1,3,5-Tris(4-carboxyphenyl)benzene (BTB) is a building block for Metal Organic Frameworks (MOFs). MOFs are 3D-microporous materials with potential applications in gas adsorption and separation technologies. BTB has been recently used as a linker to make MOFs with extremely high surface area such as MOF-177 (~ 5000 m2/g), a hydrogen absorbing material with an extremely high hydrogen storage capacity of 7.5% at 77K.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The first tritopic bridging ligand 1, 3, 5-tris (4-carboxyphenyl)-benzene (H 3 BTB) functionalized porous polyoxometalate-based metal-organic framework (POMOF): from design, synthesis to electrocatalytic properties
Dong BX, et al.
Dalton Transactions, 44(3), 1435-1440 (2015)
Chong Liu et al.
Faraday discussions, 201, 163-174 (2017-06-18)
Gradient MOFs contain directional gradients of either structure or functionality. We have successfully prepared two ternary gradient MOFs based on bMOF-100 analogues, namely bMOF-100/102/106 and bMOF-110/100/102, via cascade ligand exchange reactions. The cubic unit cell parameter discrepancy within an individual
Two-dimensional 1, 3, 5-tris (4-carboxyphenyl) benzene self-assembly at the 1-phenyloctane/graphite interface revisited
Silly F
The Journal of Physical Chemistry C, 116(18), 10029-10032 (2012)
Ram R R Prasad et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 24(57), 15309-15318 (2018-07-07)
Post-synthetic modification of the hafnium metal-organic framework MOF-808(Hf) to include triarylphosphine ligands is reported. Sulfonated phenylphosphines are incorporated without oxidation to give a "MOF ligand" that can complex late transition metals such as Ir and Rh to give a bifunctional
Meng-Yao Chao et al.
Dalton transactions (Cambridge, England : 2003), 47(38), 13722-13729 (2018-09-15)
The two-dimensional (2D) metal-organic framework (MOF) [Cd2.25Co0.75(BTB)2(DEF)4]·2(DEF)0.5 (1a, BTB = benzene-1,3,5-tribenzolate; DEF = N,N'-diethylformamide) featuring linear trimetallic cluster secondary building units (SBUs) and replaceable DEF solvates reacts smoothly with dipyridyl ligands DPS, DPDS, and BIPY (DPS = 4,4'-dipyridyl sulfide; DPDS

Articles

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service