Skip to Content
Merck
All Photos(2)

Documents

381071

Sigma-Aldrich

2-Aminoterephthalic acid

99%

Synonym(s):

2-Aminobenzene-1,4-dicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3-1,4-(CO2H)2
CAS Number:
Molecular Weight:
181.15
Beilstein:
2805625
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder, crystals or chunks

reaction suitability

reaction type: solution phase peptide synthesis

mp

324 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

Nc1cc(ccc1C(O)=O)C(O)=O

InChI

1S/C8H7NO4/c9-6-3-4(7(10)11)1-2-5(6)8(12)13/h1-3H,9H2,(H,10,11)(H,12,13)

InChI key

GPNNOCMCNFXRAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Aminoterephthalic acid can be used to synthesize:
  • Lanthanide coordination polymers with 1,10-phenanthroline by hydrothermal method.
  • Blue-emitting derivatives of 2-aminoterephthalic acid.
  • Amino-functionalized Zr-terephthalate (UiO-66), an excellent catalyst for selective synthesis of jasminaldehyde.
  • IRMOF-3, a zinc aminoterephthalate metal-organic framework useful as a catalyst for the Knoevenagel condensation of benzaldehyde and ethyl cyanoacetate.
  • Polymeric composite membrane with excellent CO2 separation capabilities.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Farhad Ahmadijokani et al.
Dalton transactions (Cambridge, England : 2003), 48(36), 13555-13566 (2019-07-30)
Mixed-matrix membranes (MMMs) are promising candidates for carbon dioxide separation. However, their application is limited due to improper dispersion of fillers within the polymer matrix, poor interaction of fillers with polymer chains, and formation of defects and micro-voids at the
Hsuan-Lan Wang et al.
ACS applied materials & interfaces, 10(11), 9332-9341 (2018-03-02)
We report an aerosol-based approach to study the thermal stability of metal-organic frameworks (MOFs) for gas-phase synthesis of MOF-based hybrid nanostructures used for highly active catalysis. Temperature-programmed electrospray-differential mobility analysis (TP-ES-DMA) provides the characterization of temperature-dependent morphological change directly in
Jianhao Qiu et al.
Chemosphere, 242, 125197-125197 (2019-11-02)
Industrial effluents often contain mixed metal ions and dyes, and it is difficult to efficiently remove both types of contaminants simultaneously. Here, MIL-125-NH2@Ag/AgCl composites were for the first time developed through a facile deposition-photoreduction method for simultaneously removing Cr(VI)/Rhodamine B
Qin Zhang et al.
Molecules (Basel, Switzerland), 23(8) (2018-08-22)
In this study, quaternary-ammonium-functionalized metal⁻organic frameworks (MOFs) Et-N-Cu(BDC-NH₂)(DMF), were prepared, characterized, and applied for the highly effective removal of metal cyanide complexes, including Pd(CN)₄²-, Co(CN)₆³-, and Fe(CN)₆³-. Batch studies were carried out, and the maximum adsorption capacities of Pd(II), Co(III)
Hao Zhang et al.
Nanomaterials (Basel, Switzerland), 10(9) (2020-09-17)
A combination of calcination and hydrothermal processing was used to prepare a g-C3N4/UiO-66-NH2/CdS photocatalyst, and the degradation of tetracycline (TC) over this material was assessed. The photocatalytic performance of this nanocomposite was approximately 4.4 and 2.3 times those of CdS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service