Skip to Content
Merck
All Photos(1)

Key Documents

759759

Sigma-Aldrich

1,3,5-Triethynylbenzene

97%

Synonym(s):

1,3,5-Triethynylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6
CAS Number:
Molecular Weight:
150.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

86-91 °C (decomposition)

SMILES string

C#Cc1cc(cc(c1)C#C)C#C

InChI

1S/C12H6/c1-4-10-7-11(5-2)9-12(6-3)8-10/h1-3,7-9H

InChI key

ZDRMMTYSQSIGRY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhao Li et al.
Environmental science & technology, 53(13), 7673-7682 (2019-06-28)
Pyrogenic carbonaceous matter (PCM) is redox-active and promotes both abiotic and biotic reactions in the environment, possibly as a result of its conductivity and phenolic/quinone functional groups. However, due to the complexity of PCM, the contribution of conductivity or phenolic/quinone
Suzhen Kong et al.
Nanoscale, 13(6), 3817-3826 (2021-02-11)
To overcome the shuttle effect in lithium-sulfur (Li-S) batteries, an sp/sp2 hybridized all-carbon interlayer by coating graphene (Gra) and hydrogen-substituted graphdiyne (HsGDY) with a specific surface area as high as 2184 m2 g-1 on a cathode is designed and prepared.
Zi Jun Wang et al.
ChemSusChem, 8(20), 3459-3464 (2015-09-10)
Metal-free visible-light photocatalysts offer a clean, sustainable solution to many pressing environmental issues. Herein, we present a molecular design strategy to fine-tune the valence and conduction band levels of a series of conjugated microporous polymer networks based on poly(benzochalcogenadiazole) for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service