Skip to Content
Merck
All Photos(2)

Documents

523763

Sigma-Aldrich

2,6-Naphthalenedicarboxylic acid

greener alternative

99%

Synonym(s):

2,6-Naphthalic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(CO2H)2
CAS Number:
Molecular Weight:
216.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>300 °C (lit.)

greener alternative category

SMILES string

OC(=O)c1ccc2cc(ccc2c1)C(O)=O

InChI

1S/C12H8O4/c13-11(14)9-3-1-7-5-10(12(15)16)4-2-8(7)6-9/h1-6H,(H,13,14)(H,15,16)

InChI key

RXOHFPCZGPKIRD-UHFFFAOYSA-N

Gene Information

human ... PTPN1(5770)

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

2,6-Naphthalenedicarboxylic acid (H2ndc) can be used in the formation of metal-organic coordination polymers (MOCPs) for potential applications in various fields such as adsorption, separation, and magnetism. It can also be used as a monomer in the production of polyesters. H2ndc can also be used in the synthesis of a metal-organic framework (MOF), which can further be used as a drug carrier.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis methods for 2, 6-naphthalenedicarboxylic acid
Elman AR
Catalysis in Industry, 1(3), 184-184 (2009)
Vladimir Setnicka et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 58(13), 2983-2989 (2002-12-13)
The host/guest complexation between cyclodextrins (CDs) and aromatic compounds was studied by vibrational circular dichroism (VCD) spectroscopy in mid-IR region. Benzoic acid, 4-aminobenzoic acid, and 2,6-naphthalene-dicarboxylic acid acting as the guests with aromatic skeleton, cause the significant changes in VCD
Structural modulation and properties of silver (I) coordination polymers from 2, 6-naphthalenedicarboxylic acid and flexible N-donor linkers
Wang XX, et al.
Transition Metal Chemistry, 40(7), 733-742 (2015)
Synthesis of metal-organic framework from iron nitrate and 2, 6-naphthalenedicarboxylic acid and its application as drug carrier
Ibrahim M, et al.
Journal of Nanoscience and Nanotechnology, 18(8), 5266-5273 (2018)
Ubed S F Arrozi et al.
Dalton transactions (Cambridge, England : 2003), 48(10), 3415-3421 (2019-02-23)
Catalytic ethylene oligomerization proceeds under mild conditions with high activity using porous or non-porous nickel-based coordination polymers/metal-organic frameworks (MOFs) as catalysts. The role of MOFs as catalyst precursors and the crucial impact of metal coordination on the catalyst activity and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service